Skip to Content
Merck
All Photos(2)

Key Documents

810192P

Avanti

16:0-06:0 NBD PS

Avanti Research - A Croda Brand 810192P, powder

Synonym(s):

1-palmitoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-phosphoserine (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€449.00

€449.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€449.00

About This Item

Empirical Formula (Hill Notation):
C34H59N6O13P
CAS Number:
Molecular Weight:
790.84
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€449.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810192P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810192P

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylserine (PS) is a membrane phospholipid enriched in the cytosolic leaflet of plasma membrane. It comprises 2-10% of total phospholipids.[1] NBD PS is a flurosently labelled phospholipid with 7-nitrobenz-2-oxa-1,3-diazol-4-yl (NBD) at the headgroup of PS.[2]

Application

16:0-06:0 NBD PS has been used as a labelled translocase to detect ATP-dependent translocation across late Golgi membranes.[3] It is also suitable for analysis of phospholipid scramblase 1 (PLSCR1) scrambling in proteoliposomes[4] and to determine enzyme specificity of lipoteichoic acid synthase enzyme (LtaS).[5]
16:0-06:0 NBD PS or 1-palmitoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-phosphoserine (ammonium salt) has been used in the 7-nitrobenz-2-oxa-1,3-diazol-4-yl (NBD) − phospholipase (PL) translocation assay to characterize an ATP-dependent flippase activity in yeast Golgi membranes.[6] It may be used: to detect vesicular stomatitis virus G (VSV G) protein mediated cell-cell fusion[7], measure the enzyme substrate specificity of lipoteichoic acid (LTA) synthase (LtaS)[8], in liposomes to determine the role of phospholipid scramblase 1 (PLSCR1) in phospholipid scrambling.[9]

Biochem/physiol Actions

Phosphatidylserine (PS) is involved in the regulation of many enzymes and structural components.[10] It serves as an essential factor in the clearance of apoptotic cells.[10] It is associated with cognitive disability in Alzheimer′s disease.[11]

Packaging

5 mL Amber Glass Screw Cap Vial (810192P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Unrecognized Function of Cholesterol: Regulating the Mechanism Controlling Membrane Phospholipid Asymmetry
Arashiki N, et al.
Biochemistry, 55(25), 3504-3504 (2016)
Osmotic stress adaptation in Lactobacillus casei BL23 leads tostructural changes in the cell wall polymer lipoteichoic acid
Palomino MM, et al.
Microbiology, 159(11), 2416-2426 (2013)
Elena Zaitseva et al.
Cell host & microbe, 22(1), 99-110 (2017-07-14)
HIV-1 entry into host cells starts with interactions between the viral envelope glycoprotein (Env) and cellular CD4 receptors and coreceptors. Previous work has suggested that efficient HIV entry also depends on intracellular signaling, but this remains controversial. Here we report
Paramasivam Natarajan et al.
Methods (San Diego, Calif.), 39(2), 163-168 (2006-07-11)
Phospholipid asymmetry is a fundamental feature of the plasma membrane of most eukaryotic cells and its regulation is linked to diverse physiological processes such as apoptosis and blood clotting [P. Williamson, R.A. Schlegel, Biochim. Biophys. Acta 1585 (2002) 53-63; R.F.
Measuring translocation of fluorescent lipid derivatives across yeast Golgi membranes
Natarajan P, et al.
Methods, 39(2), 163-168 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service