Skip to Content
Merck
All Photos(1)

Documents

890705C

Avanti

16:0-18:1 EPC (Cl Salt)

Avanti Research - A Croda Brand 890705C

Synonym(s):

1-palmitoyl-2-oleoyl-sn-glycero-3-ethylphosphocholine (chloride salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H87NO8PCl
CAS Number:
Molecular Weight:
824.59
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (890705C-10mg)
pkg of 1 × 2.5 mL (890705C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 890705C

concentration

10 mg/mL (890705C-10mg)
10 mg/mL (890705C-25mg)

lipid type

transfection
cationic lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)OCC.[Cl-]

General description

1-palmitoyl-2-oleoyl-sn-glycero-3-ethylphosphocholine (16:0-18:1 EPC) is cationic lipid containing an ethyl group attached to the phosphate moiety through an ester bond and has a choline headgroup.

Application

1-palmitoyl-2-oleoyl-sn-glycero-3-ethylphosphocholine (chloride salt) (16:0-18:1 EPC (Cl Salt)) might be used:
  • as a cationic lipid to explore the change in organization and dynamics of a membrane-bound fluorescent probe in host membranes of varying charge
  • in the preparation of small unilamellar cationic liposomes
  • in a novel albumin-associated lipoplex formulation to evaluate the antitumoral efficacy of immuno-gene therapy and “suicide” gene therapy

Biochem/physiol Actions

1-palmitoyl-2-oleoyl-sn-glycero-3-ethylphosphocholine (16:0-18:1 EPC) is a potential transfection agent, which is also like the natural lipids.

Packaging

5 mL Clear Glass Sealed Ampule (890705C-10mg)
5 mL Clear Glass Sealed Ampule (890705C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Passadouro et al.
International journal of nanomedicine, 9, 3203-3217 (2014-07-26)
Pancreatic ductal adenocarcinoma (PDAC) is a highly aggressive and mortal cancer, characterized by a set of known mutations, invasive features, and aberrant microRNA expression that have been associated with hallmark malignant properties of PDAC. The lack of effective PDAC treatment
H Faneca et al.
Biochimica et biophysica acta, 1768(5), 1093-1102 (2007-02-14)
In the present work, we used a novel albumin-associated lipoplex formulation, containing the cationic lipid 1-palmitoyl-2-oleoyl-sn-glycero-3-ethylphosphocholine (EPOPC) and cholesterol (Chol), to evaluate the antitumoral efficacy of two gene therapy strategies: immuno-gene therapy, mediated by IL-12 gene expression, and "suicide" gene
Modulation of fluorophore environment in host membranes of varying charge
Kelkar DA, et al.
Journal of Fluorescence, 13(6), 459-466 (2003)
Rumiana Koynova et al.
Biochimica et biophysica acta, 1768(2), 375-386 (2006-12-13)
Lipoplexes containing a mixture of cationic phospholipids dioleoylethylphosphatidylcholine (EDOPC) and dilauroylethylphosphatidylcholine (EDLPC) are known to be far more efficient agents in transfection of cultured primary endothelial cells than are lipoplexes containing either lipid alone. The large magnitude of the synergy
Rumiana Koynova et al.
Molecular pharmaceutics, 6(3), 951-958 (2009-04-04)
Synthetic cationic lipids are widely used components of nonviral gene carriers, and the factors regulating their transfection efficiency are the subject of considerable interest. In view of the important role that electrostatic interactions with the polyanionic nucleic acids play in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service