Skip to Content
Merck
All Photos(1)

Key Documents

890870C

Avanti

16:0 TAP

Avanti Research - A Croda Brand 890870C

Synonym(s):

1,2-dipalmitoyl-3-trimethylammonium-propane (chloride salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€181.00

€181.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€181.00

About This Item

Empirical Formula (Hill Notation):
C38H76NO4Cl
CAS Number:
Molecular Weight:
646.47
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€181.00


Check Cart for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 1 × 2.5 mL (890870C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 890870C

concentration

10 mg/mL (890870C-25mg)

lipid type

cationic lipids
transfection

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(C[N+](C)(C)C)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Cl-]

InChI

1S/C38H76NO4.ClH/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-37(40)42-35-36(34-39(3,4)5)43-38(41)33-31-29-27-25-23-21-19-17-15-13-11-9-7-2;/h36H,6-35H2,1-5H3;1H/q+1;/p-1

InChI key

KYARZEFZZOGPGS-UHFFFAOYSA-M

General description

1,2-dipalmitoyl-3-trimethylammonium-propane (16:0 TAP) is a cationic lipid and a diol based transfection reagent.

Application

1,2-dipalmitoyl-3-trimethylammonium-propane (16:0 TAP) is suitable to prepare lipid monolayer/liposome:
  • to mimic the presence of the cationic charge localized in the membrane for understanding the electrostatic effects in the biological membranes and to study its interaction with zearalenone and selenate ions[1]
  • to study its effect on the L-pro catalyzed reaction[2]
  • to serve as a positive control, to determine the effectiveness of anionic phospholipids for pro-inflammatory responses[3]

Biochem/physiol Actions

1,2-dipalmitoyl-3-trimethylammonium-propane (16:0 TAP) liposomes are known to promote elevated pro-inflammatory responses.[3]

Packaging

5 mL Clear Glass Sealed Ampule (890870C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detection of L-Proline-Catalyzed Michael Addition Reaction in Model Biomembrane
Hirose M, et al.
Journal of Chemistry, 2019 (2019)
Naomi Benne et al.
Journal of controlled release : official journal of the Controlled Release Society, 291, 135-146 (2018-10-27)
Atherosclerosis is the predominant underlying pathology of many types of cardiovascular disease and is one of the leading causes of death worldwide. It is characterized by the retention of oxidized low-density lipoprotein (ox-LDL) in lipid-rich macrophages (foam cells) in the
Barbara Gzyl-Malcher et al.
The Journal of membrane biology, 250(3), 273-284 (2017-04-30)
The impact of zearalenone and selenate ions on the monolayers of 1,2-dipalmitoyl-phosphatidylcholine (DPPC), 1,2-dipalmitoyl-3-trimethylammonium-propane (DPTAP), and the lipid mixtures (phospholipids and galactolipids) extracted from wheat plasmalemma has been studied using Langmuir trough technique and Brewster angle microscopy (BAM). The zearalenone
Ling Liu et al.
The Science of the total environment, 734, 139229-139229 (2020-05-26)
The induced membrane damage is a key mechanism for the cytotoxicity of graphene nanosheets (GNSs). In this research, the physical interaction of GNSs on model membranes was investigated using artificial membranes and plasma membrane vesicles. The effects of the GNSs
Bao Zhu et al.
Chemosphere, 247, 125876-125876 (2020-01-25)
Aluminium oxide nanoparticles (Al2O3 NPs) potentially cause health hazards after their release into the environment. The crystalline phase of Al2O3 NPs determines their surface structure and the number of functional groups. The adsorption of natural organic matter (NOM) or biomolecules

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service