Skip to Content
Merck
All Photos(1)

Documents

880610P

Avanti

14:0 PEG750 PE

Avanti Research - A Croda Brand 880610P, powder

Synonym(s):

1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-750] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 25 mg (880610P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880610P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCC)=O.[NH4+]

General description

16:0 PEG750 PE is a polyethylene glycol (PEG) conjugated phospholipid. The molecular weight and exact mass are averages based on the polydispersity of PEG.

Application

14:0 PEG750 PE has been used as a stabilizing lipid in foltate receptor-targeting magneto liposomes. It has also been used to induce modification in graphene surface.

Biochem/physiol Actions

The neutral hydrophilic polyethylene glycol (PEG) moiety in 14:0 favors efficient inhibition of protein adsorption during graphene nanopores modification.

Packaging

5 mL Clear Glass Sealed Ampule (880610P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surface modification of graphene nanopores for protein translocation
Shan YP, et al.
Nanotechnology, 24(49), 495102-495102 (2013)
Multicomponent folate-targeted magnetoliposomes: design, characterization, and cellular uptake
Bothun GD, et al.
Nanomedicine: Nanotechnology, Biology, and Medicine, 7(6), 797-805 (2011)
Y P Shan et al.
Nanotechnology, 24(49), 495102-495102 (2013-11-16)
Studies of DNA translocation through graphene nanopores have revealed their potential for DNA sequencing. Here we report a study of protein translocation through chemically modified graphene nanopores. A transmission electron microscope (TEM) was used to cut nanopores with diameters between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service