Skip to Content
Merck
All Photos(1)

Key Documents

880150P

Avanti

14:0 PEG2000 PE

Avanti Research - A Croda Brand 880150P, powder

Synonym(s):

1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-2000] (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€156.00
200 MG
€328.00
500 MG
€692.00
1 G
€1,200.00

€156.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€156.00
200 MG
€328.00
500 MG
€692.00
1 G
€1,200.00

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

€156.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 g (880150P-1g)
pkg of 1 × 200 mg (880150P-200mg)
pkg of 1 × 25 mg (880150P-25mg)
pkg of 1 × 500 mg (880150P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880150P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCC)=O.[NH4+]

InChI

1S/C125H248NO55P.H3N/c1-4-6-8-10-12-14-16-18-20-22-24-26-123(127)178-120-122(181-124(128)27-25-23-21-19-17-15-13-11-9-7-5-2)121-180-182(130,131)179-29-28-126-125(129)177-119-118-176-117-116-175-115-114-174-113-112-173-111-110-172-109-108-171-107-106-170-1

InChI key

QQHAAYKTQQJGGW-YMNXLSEASA-N

General description

14:0 PEG2000 PE, also called as 1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-2000] (DMPE-PEG2000), is a polyethylene glycol (PEG)-phospholipid conjugate. It belongs to a group of polymerizable lipids. Incorporation of DMPE-PEG in the vesicle formulation stops aggregation of vesicles upon peptide addition.[1]

Application

14:0 PEG2000 PE or 1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-2000] has been used in: giant vesicles preparation[2], lipid nanoparticle (LNP) formulation[3], unilamellar liposomes preparation for small-angle X-ray scattering (SAXS) experiments[1]

Packaging

20 mL Clear Glass Screw Cap Vial (880150P-1g)
20 mL Clear Glass Screw Cap Vial (880150P-500mg)
5 mL Amber Glass Screw Cap Vial (880150P-200mg)
5 mL Amber Glass Screw Cap Vial (880150P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Josefine Eilsø Nielsen et al.
Biochimica et biophysica acta. Biomembranes, 1861(7), 1355-1364 (2019-04-13)
The naturally occurring peptide indolicidin from bovine neutrophils exhibits strong biological activity against a broad spectrum of microorganisms. This is believed to arise from selective interactions with the negatively charged cytoplasmic lipid membrane found in bacteria. We have investigated the
L Limozin et al.
The European physical journal. E, Soft matter, 10(4), 319-330 (2004-03-12)
We studied the formation of actin scaffolds in giant vesicles of dimyristoylphosphatidylcholine (DMPC). Polymerization of actin was induced at low ionic strength through ionophore-mediated influx of Mg2+ (2 mM). The spatial organization of the filamentous actin was visualized by confocal
Pedro P G Guimaraes et al.
Journal of controlled release : official journal of the Controlled Release Society, 316, 404-417 (2019-11-05)
Messenger RNA (mRNA) has recently emerged as a promising class of nucleic acid therapy, with the potential to induce protein production to treat and prevent a range of diseases. However, the widespread use of mRNA as a therapeutic requires safe
Dominique Leboeuf et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 28(4), 1092-1104 (2020-02-24)
The N-degron pathway is an emerging target for anti-tumor therapies, because of its capacity to positively regulate many hallmarks of cancer, including angiogenesis, cell proliferation, motility, and survival. Thus, inhibition of the N-degron pathway offers the potential to be a
Abdullah Lone et al.
Molecules (Basel, Switzerland), 24(24) (2019-12-19)
The increasing emergence of multi-drug resistant bacteria is a serious threat to public health worldwide. Antimicrobial peptides have attracted attention as potential antibiotics since they are present in all multicellular organisms and act as a first line of defence against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service