Skip to Content
Merck
All Photos(1)

Key Documents

880346P

Avanti

DChemsPC

Avanti Research - A Croda Brand 880346P, powder

Synonym(s):

1,2-dicholesterylhemisuccinoyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€231.00

€231.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€231.00

About This Item

Empirical Formula (Hill Notation):
C70H116NO12P
CAS Number:
Molecular Weight:
1194.64
UNSPSC Code:
51191904
NACRES:
NA.25

€231.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 25 mg (880346P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880346P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=P([O-])(OCC[N+](C)(C)C)OC[C@H](OC(CCC(OC1CC[C@]2(C)C(C1)=CCC3C2CC[C@]4(C)C3CC[C@H]4[C@@H](C)CCCC(C)C)=O)=O)COC(CCC(OC5CC[C@]6(C)C(C5)=CCC7C6CC[C@]8(C)C7CC[C@H]8[C@@H](C)CCCC(C)C)=O)=O

InChI

1S/C70H116NO12P/c1-46(2)16-14-18-48(5)57-24-26-59-55-22-20-50-42-52(32-36-67(50,7)61(55)34-38-69(57,59)9)81-64(73)29-28-63(72)78-44-54(45-80-84(76,77)79-41-40-71(11,12)13)83-66(75)31-30-65(74)82-53-33-37-68(8)51(43-53)21-23-56-60-27-25-58(49(6)19-15-17-47

InChI key

XIHPVFHVRYJTJV-DJTREBHPSA-N

Application

DChemsPC might be used:
  • as a substitute for cholesterol in liposome formulation to study its effect on membrane stability[1]
  • in the preparation of pH-gradient liposomes[2]
  • to form sterol-phospholipid hybrid with palmitoyl-2-cholesterylhemisuccinoyl-sn-glycero-3-phosphocholine (PCholPC) and to study its properties and interactions[3]

Biochem/physiol Actions

Liposome instability in blood is caused mainly due to cholesterol exchange. Sterol modified phospholipid retains the bilayer-stabilizing effect of free cholesterol, inhibiting its transfer from the bilayer. Therefore, disterolphospholipids are ideal for liposome preparation.[4] The use of DChemsPC in liposomes prevents content leaking into the serum.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (880346P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vincent Forster et al.
Biomaterials, 33(13), 3578-3585 (2012-02-15)
Calcium channel blocker (CCB) overdose is potentially lethal. Verapamil and diltiazem are particularly prone to acute toxicity due to their dual effect on cardiac and vascular tissues. Unfortunately, conventional decontamination measures are ineffective in accelerating blood clearance and, to date
Michał Flasiński et al.
Langmuir : the ACS journal of surfaces and colloids, 32(16), 4095-4102 (2016-04-06)
The two synthetic sterol-phospholipid hybrids DCholPC and PCholPC were investigated in monolayers at the air/water interface. Study was based on π-A isotherm analysis complemented with application of grazing incidence X-ray diffraction. It was found that both compounds are capable of
Aditya G Kohli et al.
Journal of controlled release : official journal of the Controlled Release Society, 176, 86-93 (2013-12-26)
We introduce a method for tracking the rate and extent of delivery of liposome contents in vivo based on encapsulation of 4-methylumbelliferyl phosphate (MU-P), a profluorophore of 4-methylumbelliferone (MU). MU-P is rapidly dephosphorylated by endogenous phosphatases in vivo to form
Zhaohua Huang et al.
Angewandte Chemie (International ed. in English), 48(23), 4146-4149 (2009-05-09)
Extreme makeover of cholesterol: Cholesterol exchange is a major reason for the instability of liposomes in blood. The formation of a covalent hybrid between cholesterol and glycerophosphocholine preserves the bilayer-stabilizing effect of free cholesterol but prevents its transfer from the
Maryam Iman et al.
International journal of pharmaceutics, 408(1-2), 163-172 (2011-02-01)
1,2-Di-stigma-steryl-hemi-succinoyl-sn-glycero-3-phosphocholine (DSHemsPC) is a new lipid in which two molecules of stigmasterol (an inexpensive plant sterol) are covalently linked via a succinic acid to glycerophosphocholine. Since amphotericin B (AmB) interacts with sterols, we postulated that DSHemsPC could be used in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service