Skip to Content
Merck
All Photos(1)

Key Documents

880230C

Avanti

18:1 PEG5000 PE

Avanti Research - A Croda Brand 880230C

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-5000] (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€156.00

€156.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€156.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€156.00


Check Cart for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 1 × 2.5 mL (880230C-25mg)
pkg of 2 × 4 mL (880230C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880230C

concentration

10 mg/mL (880230C-25mg)
25 mg/mL (880230C-200mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC([(OCH2CH2)113OCH3])=O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+]

Application

18:1 PEG5000 PE or 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-5000 has been used in small unilamellar vesicles (SUV)[1].

Biochem/physiol Actions

18:1 PEG5000 PE, also known as 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-5000], is used along with lipid modified dendrimer copolymer to co-deliver siRNA and drugs to tumors.[2]

Packaging

5 mL Clear Glass Sealed Ampule (880230C-200mg)
5 mL Clear Glass Sealed Ampule (880230C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reconstitution of TCR signaling using supported lipid bilayers
Su X, et al.
Methods in Molecular Biology (2017)
TCR--pMHC bond conformation controls TCR ligand discrimination
Sasmal DK, et al.
Cellular & Molecular Immunology, 65-76 (2017)
Xiaolei Su et al.
Methods in molecular biology (Clifton, N.J.), 1584, 65-76 (2017-03-04)
Biochemical reconstitution has served as an important tool for understanding the mechanisms of many cellular processes including DNA replication, transcription, translation, vesicle trafficking, and ubiquitin-mediated proteolysis. Here, we demonstrate that biochemical reconstitution can be applied to studying a complex signaling
Dibyendu K Sasmal et al.
Cellular & molecular immunology, 17(3), 203-217 (2019-09-19)
A major unanswered question is how a TCR discriminates between foreign and self-peptides presented on the APC surface. Here, we used in situ fluorescence resonance energy transfer (FRET) to measure the distances of single TCR-pMHC bonds and the conformations of
Jiayi Pan et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 136, 18-28 (2019-01-12)
Multidrug resistance (MDR) significantly decreases the therapeutic efficiency of anti-cancer drugs. Its reversal could serve as a potential method to restore the chemotherapeutic efficiency. Downregulation of MDR-related proteins with a small interfering RNA (siRNA) is a promising way to reverse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service