Skip to Content
Merck
All Photos(1)

Key Documents

880230P

Avanti

18:1 PEG5000 PE

Avanti Research - A Croda Brand 880230P, powder

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-5000] (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€156.00
200 MG
€328.00

€156.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€156.00
200 MG
€328.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€156.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 2 × 100 mg (880230P-200mg)
pkg of 1 × 25 mg (880230P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880230P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC([(OCH2CH2)113OCH3])=O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+]

Application

18:1 PEG5000 PE or 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-5000 may be used in lipid bilayer preparation,[1] and DOX free base loaded mixed dendrimer micelles (MDM DOX) preparation for tumor targeted delivery of siRNA and drugs.[2]

Biochem/physiol Actions

18:1 PEG5000 PE, also referred as 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-5000] (PEG5k-DOPE) in combination with lipid modified dendrimer copolymer supports co-delivery of siRNA and drugs to tumors.[2]

Packaging

5 mL Clear Glass Sealed Ampule (880230P-200mg)
5 mL Clear Glass Sealed Ampule (880230P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TCR--pMHC bond conformation controls TCR ligand discrimination
Sasmal DK, et al.
Cellular & Molecular Immunology (2019)
Jiayi Pan et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 136, 18-28 (2019-01-12)
Multidrug resistance (MDR) significantly decreases the therapeutic efficiency of anti-cancer drugs. Its reversal could serve as a potential method to restore the chemotherapeutic efficiency. Downregulation of MDR-related proteins with a small interfering RNA (siRNA) is a promising way to reverse
Dibyendu K Sasmal et al.
Cellular & molecular immunology, 17(3), 203-217 (2019-09-19)
A major unanswered question is how a TCR discriminates between foreign and self-peptides presented on the APC surface. Here, we used in situ fluorescence resonance energy transfer (FRET) to measure the distances of single TCR-pMHC bonds and the conformations of
Xiaozheng Xu et al.
The Journal of cell biology, 219(6) (2020-05-22)
Blockade antibodies of the immunoinhibitory receptor PD-1 can stimulate the anti-tumor activity of T cells, but clinical benefit is limited to a fraction of patients. Evidence suggests that BTLA, a receptor structurally related to PD-1, may contribute to resistance to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service