Skip to Content
Merck
All Photos(1)

Key Documents

870225P

Avanti

16:0 Succinyl PE

Avanti Research - A Croda Brand 870225P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(succinyl) (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€151.00
500 MG
€873.00

€151.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€151.00
500 MG
€873.00

About This Item

Empirical Formula (Hill Notation):
C41H77NO11PNa
CAS Number:
Molecular Weight:
814.01
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€151.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 25 mg (870225P-25mg)
pkg of 1 × 500 mg (870225P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870225P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCN([H])C(CCC(O)=O)=O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C41H78NO11P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-40(46)50-35-37(36-52-54(48,49)51-34-33-42-38(43)31-32-39(44)45)53-41(47)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h37H,3-36H2,1-2H3,(H,42,43)(H,44,45)(H,48,49);/q;+1/p-1/t37-;/m1./s1

InChI key

CSBBOJBLMMDIFP-GKEJWYBXSA-M

General description

16:0 Succinyl PE, also known as 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(succinyl)(DPPE), is a head group modified functionalized lipid. Functionalized lipids act as reporter molecules.[1]

Biochem/physiol Actions

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(succinyl) (DPPE) is used in production of PHEA-EDA-DPPE-GAL copolymer. This copolymer is used in preparation of ribavirin (RBV) tripalmitate-loaded nanoparticles, that act as hepatic cell-targeted carriers for hepatitis C treatment.[2]

Packaging

20 mL Clear Glass Screw Cap Vial (870225P-500mg)
5 mL Amber Glass Screw Cap Vial (870225P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emanuela Fabiola Craparo et al.
Journal of biomedical nanotechnology, 9(6), 1107-1122 (2013-07-19)
In this paper, we describe the preparation of liver-targeted nanoparticles potentially able to carry to hepatocytes a ribavirin (RBV) prodrug, exploiting the presence of carbohydrate receptors in the liver (i.e., ASGPR in hepatocytes). These particles were obtained starting from a
Mariusz Kepczynski et al.
Biochimica et biophysica acta, 1858(10), 2362-2379 (2016-03-08)
Synthetic lipids and surfactants that do not exist in biological systems have been used for the last few decades in both basic and applied science. The most notable applications for synthetic lipids and surfactants are drug delivery, gene transfection, as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service