Skip to Content
Merck
All Photos(1)

Key Documents

870865P

Avanti

BMP22

Avanti Research - A Croda Brand, powder

Synonym(s):

4-pentadecylbenzylphosphonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€256.00
5 MG
€869.00

€256.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€256.00
5 MG
€869.00

About This Item

Empirical Formula (Hill Notation):
C22H39O3P
CAS Number:
Molecular Weight:
382.52
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€256.00


Check Cart for Availability

Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (870865P-1mg)
pkg of 1 × 5 mg (870865P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC1=CC=C(CP(O)(O)=O)C=C1

InChI key

XWVBRTAQHMKHCE-UHFFFAOYSA-N

General description

BMP (bis(monoacylglycerol)phosphate) is synthesized from phosphatidylglycerol (PG) in alveolar macrophages. BMP is a unique phospholipid having a glycerol phosphate ester at sn-1 position rather than sn-3 and has one acyl group attached to each of the glycerol moieties. The acyl groups of BMP is rich in polyunsaturated species.[1]

Biochem/physiol Actions

BMP (bis(monoacylglycerol)phosphate) (BMP) is found in the retinal pigment epithelium of healthy human retina. BMP lipids regulates cholesterol level in endosomes. It is implicated in endosomal/lysosomal storage diseases like Niemann-Pick and neuronal ceroid lipofuscinosis diseases.[2] BMP22/4PBPA is a small molecule inhibitor of autotaxin (ATX).[3][4] 4PBPA has relatively a longer half-life and also reduces the lysophosphatidic acid concentration in plasma.[3] BMP22 is implicated in reducing the bone erosion and systemic bone loss.[4]

Packaging

5ML Glass Amber Screw Cap Vial (870865P-1MG)
5ML Glass Amber Screw Cap Vial (870865P-5MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bis (monoacylglycero) phosphate lipids in the retinal pigment epithelium implicate lysosomal/endosomal dysfunction in a model of Stargardt disease and human retinas
Anderson DMG, et al.
Scientific Reports, 7(1), 17352-17352 (2017)
Osteoclast-derived Autotaxin, a distinguishing factor for inflammatory bone loss
Flammier S, et al.
Arthritis and Rheumatism (2019)
Transacylase formation of bis (monoacylglycerol) phosphate
Heravi J and Waite M
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1437(3), 277-286 (1999)
Controlling cancer through the autotaxin-lysophosphatidic acid receptor axis
Gotoh M, et al.
Biochemical Society Transactions, 40(1), 31-36 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service