Skip to Content
Merck
All Photos(1)

Key Documents

870744P

Avanti

20:5 Coenzyme A

Avanti Research - A Croda Brand 870744P, powder

Synonym(s):

(5Z,8Z,11Z,14Z,17Z-eicosapentaenoyl) Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€360.00

€360.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€360.00

About This Item

Empirical Formula (Hill Notation):
C41H73N10O17P3S
CAS Number:
Molecular Weight:
1103.06
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€360.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (870744P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870744P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C41H64N7O17P3S.3H3N/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-32(50)69-25-24-43-31(49)22-23-44-39(53)36(52)41(2,3)27-62-68(59,60)65-67(57,58)61-26-30-35(64-66(54,55)56)34(51)40(63-30)48-29-47-33-37(42)45-28-46-38(33)48;;;/h5-6,8-9,11-12,14-15,

InChI key

QCDQAUBOSVTHRQ-RWZJTUFVSA-N

Application

20:5 Coenzyme A has been used in lipidomics analysis of fatty acid-coenzyme A (FA-CoA) by liquid chromatography-mass spectrometry (LC-MS) method.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (870744P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipidomics analysis of long-chain fatty acyl-coenzyme As in liver, brain, muscle and adipose tissue by liquid chromatography/tandem mass spectrometry
Morin-Rivron D, et al.
Rapid Communications in Mass Spectrometry, 31(4), 344-350 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service