Skip to Content
Merck
All Photos(1)

Key Documents

870728P

Avanti

22:6 Coenzyme A

Avanti Research - A Croda Brand 870728P, powder

Synonym(s):

docosahexaenoyl Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€360.00

€360.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€360.00

About This Item

Empirical Formula (Hill Notation):
C43H75N10O17P3S
CAS Number:
Molecular Weight:
1129.10
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€360.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (870728P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870728P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C43H66N7O17P3S.3H3N/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-34(52)71-27-26-45-33(51)24-25-46-41(55)38(54)43(2,3)29-64-70(61,62)67-69(59,60)63-28-32-37(66-68(56,57)58)36(53)42(65-32)50-31-49-35-39(44)47-30-48-40(35)50;;;/h5-6,8-9,11-12,

InChI key

CTZVGYDJDPNALD-KEZPMSFUSA-N

General description

22: 6-Coenzyme A is synthesized from human spermatozoa.[1]

Application

22: 6-Coenzyme A has been used to determine the effects of lysophospholipid acyltransferase activities in adipocyte differentiation.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (870728P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of docosahexaenoyl coenzyme A in human spermatozoa
Jones R E and Plymate S R
Journal of Andrology, 14(6), 428-432 (1993)
Lysophosphatidylcholine acyltransferase 3 is the key enzyme for incorporating arachidonic acid into glycerophospholipids during adipocyte differentiation
Eto M, et al.
International Journal of Molecular Sciences, 13(12), 16267-16280 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service