Skip to Content
Merck
All Photos(1)

Key Documents

870704P

Avanti

04:0 Coenzyme A

Avanti Research - A Croda Brand

Synonym(s):

butanoyl Coenzyme A (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€360.00

€360.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€360.00

About This Item

Empirical Formula (Hill Notation):
C25H39N7Na3O17P3S
CAS Number:
Molecular Weight:
903.57
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€360.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (870704P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[Na+].[Na+].[Na+]

InChI

1S/C25H42N7O17P3S.3Na/c1-4-5-16(34)53-9-8-27-15(33)6-7-28-23(37)20(36)25(2,3)11-46-52(43,44)49-51(41,42)45-10-14-19(48-50(38,39)40)18(35)24(47-14)32-13-31-17-21(26)29-12-30-22(17)32;;;/h12-14,18-20,24,35-36H,4-11H2,1-3H3,(H,27,33)(H,28,37)(H,41,42)(H,43,44)(H2,26,29,30)(H2,38,39,40);;;/q;3*+1/p-3/t14-,18?,19+,20+,24-;;;/m1.../s1

InChI key

PRMIKLPNYYDDCC-XRHOYZRHSA-K

General description

04:0 Coenzyme A, also known as butanoyl coenzyme A, is a coenzyme A derivative of butanoic acid. 04:0 Coenzyme A is a short-chain acyl CoA[1] and is an intermediate of fatty acid degradation.[2]

Biochem/physiol Actions

04:0 Coenzyme A acts as a source of N-acyl chain in the synthesis of N-butanoyl-L-homoserine lactone (BHL) by LuxI homologue RhlI (VsmI).[2] Butanoyl coenzyme A serves as an intermediate in the synthesis of butyrate, which is involved in maintaining colonic homeostasis and gut health.[3] It also has an ability to inhibit citrate synthase (CS).[1] 

Packaging

5 mL Amber Glass Screw Cap Vial (870704P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julian Trachsel et al.
Applied and environmental microbiology, 82(22), 6788-6798 (2016-10-30)
Studying the host-associated butyrate-producing bacterial community is important, because butyrate is essential for colonic homeostasis and gut health. Previous research has identified the butyryl coenzyme A (CoA):acetate-CoA transferase (EC 2.3.8.3) as a gene of primary importance for butyrate production in
Y Jiang et al.
Molecular microbiology, 28(1), 193-203 (1998-05-21)
In Pseudomonas aeruginosa, synthesis of the quorum-sensing signal molecules N-butanoyl-L-homoserine lactone (BHL) and N-hexanoyl-L-homoserine lactone (HHL) requires the Luxl homologue Rhll(Vsml). By using thin-layer chromatography in conjunction with high-performance liquid chromatography (HPLC) and mass spectrometry, we show that purified Rhll
J C Lai et al.
Research communications in chemical pathology and pharmacology, 82(3), 331-338 (1993-12-01)
We investigated the hypothesis that one mechanism underlying fatty acid toxicity is the selective inhibition of rate-limiting and/or regulated tricarboxylic acid cycle and related enzymes by fatty acyl coenzyme A (CoA) derivatives by examining the effects of several fatty acyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service