Skip to Content
Merck
All Photos(1)

Key Documents

870717P

Avanti

17:0 Coenzyme A

Avanti Research - A Croda Brand 870717P, powder

Synonym(s):

heptadecanoyl Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€360.00

€360.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€360.00

About This Item

Empirical Formula (Hill Notation):
C38H77N10O17P3S
CAS Number:
Molecular Weight:
1071.06
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€360.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (870717P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870717P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCCCCCCCCCCCCCCC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C38H68N7O17P3S/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-29(47)66-22-21-40-28(46)19-20-41-36(50)33(49)38(2,3)24-59-65(56,57)62-64(54,55)58-23-27-32(61-63(51,52)53)31(48)37(60-27)45-26-44-30-34(39)42-25-43-35(30)45/h25-27,31-33,37,48-49H,4-24H2,1-3H3,(H,40,46)(H,41,50)(H,54,55)(H,56,57)(H2,39,42,43)(H2,51,52,53)/t27-,31-,32-,33+,37-/m1/s1

InChI key

DRABUZIHHACUPI-DUPKZGIXSA-N

Application

17:0 Coenzyme A or heptadecanoyl Coenzyme A (ammonium salt) has been used as an internal standard for long-chain acyl-CoA (LCACoA) analysis by liquid chromatography/tandem mass spectrometry.[1][2] It has also been used as an internal standard to measure the percentage recovery of fatty acyl-CoAs by high performance liquid chromatography.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (870717P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas A Bell et al.
Journal of lipid research, 48(5), 1122-1131 (2007-02-06)
ACAT2, the enzyme responsible for the formation of cholesteryl esters incorporated into apolipoprotein B-containing lipoproteins by the small intestine and liver, forms predominantly cholesteryl oleate from acyl-CoA and free cholesterol. The accumulation of cholesteryl oleate in plasma lipoproteins has been
Matías Cabruja et al.
Applied microbiology and biotechnology, 100(16), 7239-7248 (2016-06-09)
Acyl-CoAs are crucial compounds involved in essential metabolic pathways such as the Krebs cycle and lipid, carbohydrate, and amino acid metabolisms, and they are also key signal molecules involved in the transcriptional regulation of lipid biosynthesis in many organisms. In
Agnieszka U Blachnio-Zabielska et al.
Rapid communications in mass spectrometry : RCM, 25(15), 2223-2230 (2011-07-08)
Long-chain acyl-coenzymes A (acyl-CoAs) (LCACoA) are the activated forms of long-chain fatty acids and serve as key lipid metabolites. Excess accumulation of intracellular LCACoA, diacylglycerols (DAGs) and ceramides may create insulin resistance with respect to glucose metabolism. We present a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service