Skip to Content
Merck
All Photos(1)

Key Documents

860822P

Avanti

22:0(2S-OH) Ceramide

Avanti Research - A Croda Brand 860822P, powder

Synonym(s):

N-(2′-(S)-hydroxybehenoyl)-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€299.00

€299.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€299.00

About This Item

Empirical Formula (Hill Notation):
C40H79NO4
CAS Number:
Molecular Weight:
638.06
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€299.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (860822P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860822P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC/C=C/[C@@H](O)[C@@H](NC([C@@H](O)CCCCCCCCCCCCCCCCCCCC)=O)CO

InChI

1S/C40H79NO4/c1-3-5-7-9-11-13-15-17-18-19-20-21-23-25-27-29-31-33-35-39(44)40(45)41-37(36-42)38(43)34-32-30-28-26-24-22-16-14-12-10-8-6-4-2/h32,34,37-39,42-44H,3-31,33,35-36H2,1-2H3,(H,41,45)/b34-32+/t37-,38+,39-/m0/s1

InChI key

FCCZDEAAQZWMGI-RHJYVNCBSA-N

General description

Ceramides containing 2-hydroxy fatty acids (hFA) are primarily found in the epidermis.[1] 22:0(2S-OH) Ceramide is a 2S hydroxylated behenic acid-containing sphingolipid.[2]

Biochem/physiol Actions

Sphingolipids containing hydroxy fatty acid (hFA) are implicated in cell signaling,[3] cell differentiation and apoptosis.[2] These lipids, which are found majorly in myelin sheath helps in formation and function of myelin.[4] NAD(P)H-dependent enzyme, fatty acid 2-hydroxylase (FA2H) catalyzes the synthesis of hFA-ceramide.[5] hFA-ceramides aid in epidermal permeability barrier function.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (860822P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Absence of 2-hydroxylated sphingolipids is compatible with normal neural development but causes late-onset axon and myelin sheath degeneration
Zoller I, et al.
The Journal of Neuroscience, 28(39), 9741-9754 (2008)
Inge Zöller et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 28(39), 9741-9754 (2008-09-26)
Sphingolipids containing 2-hydroxylated fatty acids are among the most abundant lipid components of the myelin sheath and therefore are thought to play an important role in formation and function of myelin. To prove this hypothesis, we generated mice lacking a
Stereospecificity of fatty acid 2-hydroxylase and differential functions of 2-hydroxy fatty acid enantiomers
Guo L, et al.
Journal of Lipid Research, 53(7), 1327-1335 (2012)
Normal fur development and sebum production depends on fatty acid 2-hydroxylase expression in sebaceous glands
Maier H, et al.
The Journal of Biological Chemistry, 286(29), 25922-25934 (2011)
Fatty acid 2-Hydroxylation in mammalian sphingolipid biology
Hama H, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1801(4), 405-414 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service