Skip to Content
Merck
All Photos(2)

Key Documents

860103P

Avanti

18:1 Phosphatidylpropanol

1,2-dioleoyl-sn-glycero-3-phosphopropanol (sodium salt), powder

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-phosphopropanol (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€135.00

€135.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€135.00

About This Item

Empirical Formula (Hill Notation):
C42H78O8PNa
CAS Number:
Molecular Weight:
765.03
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€135.00


Estimated to ship on24 May 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (860103P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860103P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCC)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI

1S/C42H79O8P.Na/c1-4-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-41(43)47-38-40(39-49-51(45,46)48-37-6-3)50-42(44)36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-5-2;/h19-22,40H,4-18,23-39H2,1-3H3,(H,45,46);/q;+1/p-1/b21-19-,22-20-;/t40-;/m1./s1

InChI key

ZGATVCGNENIEOO-MYJLHYLNSA-M

Application

18:1 Phosphatidylpropanol or 1,2-dioleoyl-sn-glycero-3-phosphopropanol (sodium salt) may be used as an internal standard:
  • in ultra-high performance liquid chromatography tandem mass spectrometry (UHPLC-MS/MS) to evaluate its matrix effect in blood samples for the validation of phosphatidylethanol (PEth)[1]
  • for the quantification of PEth using liquid chromatography tandem mass spectrometry (LC-MS/MS) in human blood[2]
  • for the quantification of PEth using liquid chromatography electrospray ionization tandem mass spectrometric (LC/ESI-MS/MS) in human blood[3]

Packaging

5 mL Clear Glass Sealed Ampule (860103P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yufang Zheng et al.
Clinica chimica acta; international journal of clinical chemistry, 412(15-16), 1428-1435 (2011-05-03)
Phosphatidylethanol (PEth) is a group of phospholipids formed from ethanol and phosphatidylcholine by action of phospholipase D. Measurement of PEth in whole blood samples is employed as an alcohol biomarker. This work aimed to further develop an LC-MS method for
Xinyu Zhang et al.
Analytica chimica acta, 963, 68-75 (2017-03-25)
The importance of ethanol non-oxidative metabolites as the specific biomarkers of alcohol consumption in clinical and forensic settings is increasingly acknowledged. Simultaneous determination of these metabolites can provide a wealth of information like drinking habit and history, but it was
Natalie Kummer et al.
Analytical and bioanalytical chemistry, 408(3), 825-838 (2015-11-26)
Phosphatidylethanol species (PEths) are promising biomarkers of alcohol consumption. Here, we report on the set-up, validation, and application of a novel UHPLC-ESI-MS/MS method for the quantification of PEth 16:0/18:1, PEth 18:1/18:1, and PEth 16:0/16:0 in whole blood (30 μL) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service