Skip to Content
Merck
All Photos(1)

Key Documents

860529P

Avanti

C12 Sphingosyl PE (d17:1/12:0)

Avanti Research - A Croda Brand 860529P, powder

Synonym(s):

N-lauroyl-D-erythro-sphingosyl phosphoethanolamine (C17 base)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€436.00

€436.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€436.00

About This Item

Empirical Formula (Hill Notation):
C31H63N2O6P
CAS Number:
Molecular Weight:
590.82
UNSPSC Code:
12352211
NACRES:
NA.25

€436.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (860529P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860529P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCC/C=C/[C@@]([H])(O)[C@@]([H])(NC(CCCCCCCCCCC)=O)COP([O-])(OCC[NH3+])=O

InChI

1S/C31H63N2O6P/c1-3-5-7-9-11-13-14-15-17-18-20-22-24-30(34)29(28-39-40(36,37)38-27-26-32)33-31(35)25-23-21-19-16-12-10-8-6-4-2/h22,24,29-30,34H,3-21,23,25-28,32H2,1-2H3,(H,33,35)(H,36,37)/b24-22+/t29-,30+/m0/s1

InChI key

ZIEWVPJUBVRCAL-DQBIDPIGSA-N

General description

C12 Sphingosyl PE (d17:1/12:0) or N-lauroyl-D-erythro-sphingosyl phosphoethanolamine (C17 base) is a synthetic phosphosphingolipid. Sphingosyl phosphoethanolamine (PE) is a derivative of sphingosine, wherein C-2 amine and C-1 hydroxyl groups of sphingosine backbone are substituted with lauric acid and PE groups, respectively.

Application

C12 Sphingosyl PE (d17:1/12:0) or N-lauroyl-D-erythro-sphingosyl phosphoethanolamine (C17 base) may be used:
  • as a lipid internal standard for Folch lipid extraction[1]
  • as an internal standard for the extraction of lipids using Bligh and Dyer method[2]
  • used as a standard to quantify ceramide phosphoethanolamine (CerPE) lipids present in Bacteroides thetaiotaomicron[3]

Packaging

5 mL Amber Glass Screw Cap Vial (860529P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ion Mobility-Mass Spectrometry (2019)
Eric M Brown et al.
Cell host & microbe, 25(5), 668-680 (2019-05-10)
Sphingolipids are structural membrane components and important eukaryotic signaling molecules. Sphingolipids regulate inflammation and immunity and were recently identified as the most differentially abundant metabolite in stool from inflammatory bowel disease (IBD) patients. Commensal bacteria from the Bacteroidetes phylum also
Hema Balakrishna Bhat et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(9), 3920-3934 (2015-06-11)
Ceramide phosphoethanolamine (CPE), a sphingomyelin analog, is a major sphingolipid in invertebrates and parasites, whereas only trace amounts are present in mammalian cells. In this study, mushroom-derived proteins of the aegerolysin family—pleurotolysin A2 (PlyA2; K(D) = 12 nM), ostreolysin (Oly;

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service