Skip to Content
Merck
All Photos(1)

Key Documents

860500C

Avanti

C16 Cyanine 5 SM

chloroform solution, Avanti Research - A Croda Brand

Synonym(s):

N-palmitoyl sphingomyelin-N-(Cyanine 5)

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
€274.00

€274.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 μG
€274.00

About This Item

Empirical Formula (Hill Notation):
C72H119ClN5O7P
CAS Number:
Molecular Weight:
1233.17
UNSPSC Code:
12352211
NACRES:
NA.25

€274.00


Check Cart for Availability

Request a Bulk Order

Assay

99% (TLC)

form

chloroform solution

packaging

package of 1 × 250 μg

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

Packaging

5 mL Amber Glass Screw Cap Vial (860500C-250UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service