Skip to Content
Merck
All Photos(1)

Key Documents

810337C

Avanti

18:1 Cyanine 7 PE

Avanti Research - A Croda Brand 810337C

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(Cyanine 7)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C78H124ClN4O9P
CAS Number:
Molecular Weight:
1328.27
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810337C-1mg)
pkg of 5 × 1 mL (810337C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810337C

concentration

1 mg/mL (810337C-1mg)
1 mg/mL (810337C-5mg)

shipped in

dry ice

storage temp.

−20°C

General description

18:1 Cy7 PE or 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(Cyanine 7) is a phosphoethanolamine (PE) derivative wherein the amine and hydroxyl groups of PE are substituted with cyanine 7 and dioleoyl-sn-glycerol.

Application

18:1 Cy7 PE or 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(Cyanine 7) has been used in Forster resonance energy transfer (FRET) dye pairs for small unilamellar vesicles (SUVs) preparation to serve as a model cellular system and to study membrane tension through subjection to osmotic pressure.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (810337C-1mg)
5 mL Amber Glass Screw Cap Vial (810337C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margrethe A Boyd et al.
Biophysical journal, 115(7), 1307-1315 (2018-09-17)
Cells dynamically regulate their membrane surface area during a variety of processes critical to their survival. Recent studies with model membranes have pointed to a general mechanism for surface area regulation under tension in which cell membranes unfold or take

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service