Skip to Content
Merck
All Photos(2)

Key Documents

850549P

Avanti

DORI

>99% (TLC), Avanti Research - A Croda Brand

Synonym(s):

N-(2-hydroxyethyl)-N,N-dimethyl-2,3-bis(oleoyloxy)propan-1-aminium bromide

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€327.00

€327.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€327.00

About This Item

Empirical Formula (Hill Notation):
C43H82BrNO5
Molecular Weight:
773.02
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€327.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

package of 1 × 25 mg ((840531P-25
(840531P-25MG))

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

advanced drug delivery

lipid type

cationic lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(C[N+](C)(C)CCO)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Br-]

InChI key

NNIFGWCTDFSLJC-JDVCJPALSA-M

Packaging

5 mL Amber Glass Screw Cap Vial (850549P-25MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service