Skip to Content
Merck
All Photos(1)

Key Documents

890898P

Avanti

DOTMA

Avanti Research - A Croda Brand

Synonym(s):

1,2-di-O-octadecenyl-3-trimethylammonium propane (chloride salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€322.00
200 MG
€652.00

€322.00


Estimated to ship on28 March 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€322.00
200 MG
€652.00

About This Item

Empirical Formula (Hill Notation):
C42H84ClNO2
CAS Number:
Molecular Weight:
670.57
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€322.00


Estimated to ship on28 March 2025


Request a Bulk Order

form

powder

packaging

pkg of 2 × 100 mg (890898P-200mg)
pkg of 1 × 25 mg (890898P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

advanced drug delivery

lipid type

cationic lipids
transfection

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(C[N+](C)(C)C)(OCCCCCCCC/C=C\CCCCCCCC)COCCCCCCCC/C=C\CCCCCCCC.[Cl-]

Looking for similar products? Visit Product Comparison Guide

Application

DOTMA is suitable for use as a building block of miRNA delivery system and in the preparation.[1]

Biochem/physiol Actions

DOTMA is a cationic lipid, used as a non-viral vector for gene therapy. It exhibits effective in vitro and in vivo gene transfection. DOTMA induces a positive charge on the liposomes and thus promotes efficient liposome - cell membrane interaction.[1]

Packaging

5 mL Clear Glass Sealed Ampule (890898P-200mg)
5 mL Clear Glass Sealed Ampule (890898P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The use of cationic microbubbles to improve ultrasound-targeted gene delivery to the ischemic myocardium
Sun L, et al.
Biomaterials, 34(8), 2107-2116 (2013)
Yun Wu et al.
Molecular therapy. Nucleic acids, 2, e84-e84 (2013-04-18)
MicroRNA-29b (miR-29b) expression has been shown to be reduced in non-small-cell lung cancer (NSCLC) tissues. Here, we have identified the oncogene cyclin-dependent protein kinase 6 (CDK6) as a direct target of miR-29b in lung cancer. We hypothesized that in vivo
Jinhong Meng et al.
Scientific reports, 10(1), 1046-1046 (2020-01-25)
P53 mutations are responsible for drug-resistance of tumour cells which impacts on the efficacy of treatment. Alternative tumour suppressor pathways need to be explored to treat p53- deficient tumours. The E3 ubiquitin ligase, ITCH, negatively regulates the tumour suppressor protein
Jayesh A Kulkarni et al.
ACS nano, 12(5), 4787-4795 (2018-04-04)
Lipid nanoparticles (LNPs) containing short interfering RNA (LNP-siRNA) and optimized ionizable cationic lipids are now clinically validated systems for silencing disease-causing genes in hepatocytes following intravenous administration. However, the mechanism of formation and certain structural features of LNP-siRNA remain obscure.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service