Skip to Content
Merck
All Photos(2)

Key Documents

850545P

Avanti

LDAO

Avanti Research - A Croda Brand

Synonym(s):

N,N-dimethyldodecylamine N-oxide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€77.20
5 G
€309.00

€77.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€77.20
5 G
€309.00

About This Item

Empirical Formula (Hill Notation):
C14H31NO
CAS Number:
Molecular Weight:
229.40
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.25

€77.20


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

mol wt

229.40 g/mol

packaging

pkg of 1 × 1 g (850545P-1g)
pkg of 1 × 5 g (850545P-5g)

manufacturer/tradename

Avanti Research - A Croda Brand

technique(s)

electrophoresis: suitable

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCC[N+](C)([O-])C

InChI

1S/C14H31NO/c1-4-5-6-7-8-9-10-11-12-13-14-15(2,3)16/h4-14H2,1-3H3

InChI key

SYELZBGXAIXKHU-UHFFFAOYSA-N

General description

An optimized reverse micelle surfactant system has been developed for solution nuclear magnetic resonance studies of encapsulated proteins and nucleic acids dissolved in low viscosity fluids. Comprising the nonionic 1-decanoyl-rac-glycerol and the zwitterionic lauryldimethylamine-N-oxide (10MAG/LDAO), this mixture is shown to efficiently encapsulate a diverse set of proteins and nucleic acids. Chemical shift analyses of these systems show that high structural fidelity is achieved upon encapsulation.
LDAO (N,N-Dimethyldodecylamine N-oxide) is a zwitterionic detergent.[1]

Application

LDAO is suitable to use in crystallization of membrane proteins.[1] It is also suitable to enhance the detection of high molecular weight proteins.[2]

Biochem/physiol Actions

LDAO can be used to disrupt phospholipid bilayer of cells.[1]

Packaging

20 mL Clear Glass Screw Cap Vial (850545P-1g)
60 mL Amber Wide Mouth Glass Bottle with Screw Cap (850545P-5g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detergents for the stabilization and crystallization of membrane proteins
Prive GG
Methods, 41(4), 388-397 (2007)
MALDI mass spectrometry analysis of high molecular weight proteins from whole bacterial cells: pretreatment of samples with surfactants
Meetani MA and Voorhees KJ
Journal of the American Society For Mass Spectrometry, 16(9), 1422-1426 (2005)
Igor Dodevski et al.
Journal of the American Chemical Society, 136(9), 3465-3474 (2014-02-06)
An optimized reverse micelle surfactant system has been developed for solution nuclear magnetic resonance studies of encapsulated proteins and nucleic acids dissolved in low viscosity fluids. Comprising the nonionic 1-decanoyl-rac-glycerol and the zwitterionic lauryldimethylamine-N-oxide (10MAG/LDAO), this mixture is shown to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service