Skip to Content
Merck
All Photos(2)

Key Documents

850521P

Avanti

n-decyl-β-D-maltoside

n-decyl-β-D-maltopyranoside, powder

Synonym(s):

DM; decyl 4-O-α-D-glucopyranosyl-β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€118.00

€118.00


Estimated to ship on09 April 2025


Request a Bulk Order

Select a Size

Change View
1 G
€118.00

About This Item

Empirical Formula (Hill Notation):
C22H42O11
CAS Number:
Molecular Weight:
482.56
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.25

€118.00


Estimated to ship on09 April 2025


Request a Bulk Order

Assay

>99% (contains <2% alpha isomer, TLC)

form

powder

mol wt

482.56 g/mol

packaging

pkg of 1 × 1 g (850521P-1g)

manufacturer/tradename

Avanti Research - A Croda Brand 850521P

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](OCCCCCCCCCC)[C@H](O)[C@@H](O)[C@@H]1O[C@@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O2

InChI

1S/C22H42O11/c1-2-3-4-5-6-7-8-9-10-30-21-19(29)17(27)20(14(12-24)32-21)33-22-18(28)16(26)15(25)13(11-23)31-22/h13-29H,2-12H2,1H3/t13-,14-,15-,16+,17-,18-,19-,20-,21-,22-/m1/s1

InChI key

WOQQAWHSKSSAGF-WXFJLFHKSA-N

Application

n-decyl-β-D-maltoside may be used as a detergent component in decyl maltopyranoside (DM)/lysis buffer to prevent protein aggregation during purification.[1] It may also be used as a component of lysis buffer in serum amyloid A protein purification.[2]

Packaging

20 mL Clear Glass Screw Cap Vial (850521P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zehan Hu et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(38), 19077-19082 (2019-09-06)
Serum amyloid A (SAA) proteins are strongly induced in the liver by systemic infection and in the intestine by bacterial colonization. In infected mice, SAA proteins circulate in association with the vitamin A derivative retinol, suggesting that SAAs transport retinol
June-Yong Lee et al.
Cell, 180(1), 79-91 (2019-12-24)
Lymphoid cells that produce interleukin (IL)-17 cytokines protect barrier tissues from pathogenic microbes but are also prominent effectors of inflammation and autoimmune disease. T helper 17 (Th17) cells, defined by RORγt-dependent production of IL-17A and IL-17F, exert homeostatic functions in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service