Skip to Content
Merck
All Photos(2)

Key Documents

850480C

Avanti

16:0-18:0 (6-7BR) PC

1-palmitoyl-2-(6,7-dibromo)stearoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-hexadecanoyl-2-(6,7-dibromooctadecanoyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€455.00

€455.00


Estimated to ship on08 April 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€455.00

About This Item

Empirical Formula (Hill Notation):
C42H82NO8PBr2
CAS Number:
Molecular Weight:
919.88
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€455.00


Estimated to ship on08 April 2025


Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (850480C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850480C

concentration

10 mg/mL (850480C-10mg)

lipid type

omega FAs
phospholipids

shipped in

dry ice

storage temp.

−20°C

General description

16:0-18:0 (6-7BR) PC (1-palmitoyl-2-(6,7-dibromo) stearoyl-sn-glycero-3-phosphocholine) is a brominated-PC lipid.[1]

Application

16:0-18:0 (6-7BR) PC (1-palmitoyl-2-(6,7-dibromo) stearoyl-sn-glycero-3-phosphocholine) may be used:
  • to prepare large unilamellar liposomes for parallax analysis[2]
  • in phospholipid vesicle preparation for tryptophan (Trp) penetration depth experiments[1]
  • to study the effect of bromination at different positions along the fatty acid chains[3]

Biochem/physiol Actions

16:0-18:0 (6-7BR) PC (1-palmitoyl-2-(6,7-dibromo) stearoyl-sn-glycero-3-phosphocholine) serves as a shallow quencher.[2]

Packaging

5 mL Clear Glass Sealed Ampule (850480C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tim Rasmussen et al.
Journal of molecular biology, 431(17), 3339-3352 (2019-06-08)
All membrane proteins have dynamic and intimate relationships with the lipids of the bilayer that may determine their activity. Mechanosensitive channels sense tension through their interaction with the lipids of the membrane. We have proposed a mechanism for the bacterial
Johanna C Karst et al.
The Journal of biological chemistry, 287(12), 9200-9212 (2012-01-14)
The adenylate cyclase (CyaA) toxin, one of the virulence factors secreted by Bordetella pertussis, the pathogenic bacteria responsible for whooping cough, plays a critical role in the early stages of respiratory tract colonization by this bacterium. The CyaA toxin is
John M Harrington et al.
PloS one, 7(9), e44384-e44384 (2012-09-13)
The bloodstream developmental forms of pathogenic African trypanosomes are uniquely susceptible to killing by small hydrophobic peptides. Trypanocidal activity is conferred by peptide hydrophobicity and charge distribution and results from increased rigidity of the plasma membrane. Structural analysis of lipid-associated
Candace M Pfefferkorn et al.
Biochimica et biophysica acta, 1848(11 Pt A), 2821-2828 (2015-08-25)
Apolipoproteins are essential human proteins for lipid metabolism. Together with phospholipids, they constitute lipoproteins, nm to μm sized particles responsible for transporting cholesterol and triglycerides throughout the body. To investigate specific protein-lipid interactions, we produced and characterized three single-Trp containing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service