Merck
  • Tryptophan probes reveal residue-specific phospholipid interactions of apolipoprotein C-III.
Tryptophan probes reveal residue-specific phospholipid interactions of apolipoprotein C-III.
Biochimica et biophysica acta (2015-08-25)
Candace M Pfefferkorn, Robert L Walker, Yi He, James M Gruschus, Jennifer C Lee
ABSTRACT
Apolipoproteins are essential human proteins for lipid metabolism. Together with phospholipids, they constitute lipoproteins, nm to μm sized particles responsible for transporting cholesterol and triglycerides throughout the body. To investigate specific protein-lipid interactions, we produced and characterized three single-Trp containing apolipoprotein C-III (ApoCIII) variants (W42 (W54F/W65F), W54 (W42F/W65F), W65 (W42F/W54F)). Upon binding to phospholipid vesicles, wild-type ApoCIII adopts an α-helical conformation (50% helicity) as determined by circular dichroism spectroscopy with an approximate apparent partition constant of 3×10(4) M(-1). Steady-state and time-resolved fluorescence measurements reveal distinct residue-specific behaviors with W54 experiencing the most hydrophobic environment followed by W42 and W65. Interestingly, time-resolved anisotropy measurements show a converse trend for relative Trp mobility with position 54 being the least immobile. To determine the relative insertion depths of W42, W54, and W65 in the bilayer, fluorescence quenching experiments were performed using three different brominated lipids. W65 had a clear preference for residing near the headgroup while W54 and W42 sample the range of depths ~8-11 Å from the bilayer center. On average, W54 is slightly more embedded than W42. Based on Trp spectral differences between ApoCIII binding to phospholipid vesicles and sodium dodecyl sulfate micelles, we suggest that ApoCIII adopts an alternate helical conformation on the bilayer which could have functional implications.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Avanti
16:0-18:0 (9-10BR) PC, 1-palmitoyl-2-(9,10-dibromo)stearoyl-sn-glycero-3-phosphocholine, chloroform
Avanti
16:0-18:0 (6-7BR) PC, 1-palmitoyl-2-(6,7-dibromo)stearoyl-sn-glycero-3-phosphocholine, chloroform
SAFC
Tromethamine
Sigma-Aldrich
N-Acetyl-L-tryptophanamide
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Avanti
16:0-18:0 (11-12BR) PC, 1-palmitoyl-2-(11,12-dibromo)stearoyl-sn-glycero-3-phosphocholine, powder