Skip to Content
Merck
All Photos(2)

Key Documents

850368C

Avanti

20:0 PC

1,2-diarachidoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1,2-dieicosanoyl-sn-glycero-3-phosphocholine; PC(20:0/20:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€102.00

€102.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€102.00

About This Item

Empirical Formula (Hill Notation):
C48H96NO8P
CAS Number:
Molecular Weight:
846.25
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€102.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850368C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850368C

concentration

10 mg/mL (850368C-25mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCCCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C48H96NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-38-40-47(50)54-44-46(45-56-58(52,53)55-43-42-49(3,4)5)57-48(51)41-39-37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h46H,6-45H2,1-5H3/t46-/m1/s1

InChI key

YKIOPDIXYAUOFN-YACUFSJGSA-N

General description

The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.
The mucosal layer of the colon has phosphatidylcholine (PC) as a vital constituent.[1]

Application

20:0 PC has been used in collision cross section (CCS) calibration mixture for lipid identification.[2]
It may be used:
  • as a component in giant unilamellar vesicles (GUVs) for a two-photon fluorescence microscopy study[3]
  • to prepare small unilamellar vesicles (SUVs) to study the effect of hydrophobic mismatch on gramicidin conformation and organization[4]
  • as a component in giant unilamellar vesicles (GUVs) to study its interaction with Crotalus atrox-secreted phospholipase A2 (sPLA2)[5]

Biochem/physiol Actions

Phosphatidylcholine (PC) may participate in the signaling of neuronal differentiation.[6] It can serve as a surfactant to block the penetrance of bacteria by generating a hydrophobic surface.[1]

Packaging

5 mL Clear Glass Sealed Ampule (850368C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Use of LipidIMMS Analyzer for Lipid Identification in Ion Mobility-Mass Spectrometry-Based Untargeted Lipidomics
Ion Mobility-Mass Spectrometry, 269-282 (2020)
Inflammatory Bowel Disease
Integrative Medicine, 501-516 (2018)
Susana A Sanchez et al.
Biophysical journal, 82(4), 2232-2243 (2002-03-28)
We describe the interaction of Crotalus atrox-secreted phospholipase A2 (sPLA2) with giant unilamellar vesicles (GUVs) composed of single and binary phospholipid mixtures visualized through two-photon excitation fluorescent microscopy. The GUV lipid compositions that we examined included 1-palmitoyl-2-oleoyl-phosphatidylcholine, 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC), and
L A Bagatolli et al.
Biophysical journal, 79(1), 434-447 (2000-06-27)
Giant unilamellar vesicles (GUVs) composed of different phospholipid binary mixtures were studied at different temperatures, by a method combining the sectioning capability of the two-photon excitation fluorescence microscope and the partition and spectral properties of 6-dodecanoyl-2-dimethylamino-naphthalene (Laurdan) and Lissamine rhodamine
Devaki A Kelkar et al.
Biochimica et biophysica acta, 1768(5), 1103-1113 (2007-02-27)
The matching of hydrophobic lengths of integral membrane proteins and the surrounding lipid bilayer is an important factor that influences both structure and function of integral membrane proteins. The ion channel gramicidin is known to be uniquely sensitive to membrane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service