Skip to Content
Merck

Skip To

8.03452

Dichlorodimethylsilane

for synthesis

Synonym(s):

Dichlorodimethylsilane, Dimethyldichlorosilane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
250 mL
Check Cart for Availability
€ 44,70
1 L
Check Cart for Availability
€ 129,00

About This Item

Linear Formula:
(CH3)2SiCl2
CAS Number:
Molecular Weight:
129.06
UNSPSC Code:
12352103
EC Index Number:
200-901-0
NACRES:
NA.22
MDL number:
Form:
liquid

€ 44,70


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

<200 hPa ( 20 °C)

Quality Segment

form

liquid

autoignition temp.

425 °C

potency

6056 mg/kg LD50, oral (Rat)

expl. lim.

1.75-48.5 % (v/v)

bp

70 °C/1013 hPa

mp

-76 °C

transition temp

flash point -5 °C

density

1.07 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

[Si](Cl)(Cl)(C)C

InChI

1S/C2H6Cl2Si/c1-5(2,3)4/h1-2H3

InChI key

LIKFHECYJZWXFJ-UHFFFAOYSA-N

Application



  • Modification of silica nanoparticles by dichlorodimethylsilane for foam stability and decontamination efficiency: This study explored the surface modification of silica nanoparticles using dichlorodimethylsilane to enhance foam stability and decontamination efficiency (IH Yoon et al., 2016 - kns.org). View Article.


  • Formation of a macrocycle from dichlorodimethylsilane and a pyridoxalimine Schiff base ligand: Researchers reported the synthesis of a novel macrocyclic silicon compound through the reaction of dichlorodimethylsilane with a Schiff base ligand (U Böhme et al., 2021 - scripts.iucr.org). View Article.


  • Use of Dichlorodimethylsilane to Produce Polydimethylsiloxane as a Substitute for Vitreous Humour: Characteristics and In Vitro Toxicity: This research evaluated polydimethylsiloxane produced from dichlorodimethylsilane as a substitute for vitreous humour in vitreoretinal surgery (DG Auliya et al., 2023 - mdpi.com). View Article.


  • Dichlorodimethylsilane mediated one-step synthesis of hydrophilic and hydrophobic silica nanoparticles: The study focused on the use of dichlorodimethylsilane for the one-step synthesis of silica nanoparticles, examining the effects of various synthesis parameters (F Ebrahimi et al., 2017 - Elsevier). View Article.


  • Hydrophobization of cotton fabric with silanes with different substituents: This paper explored the hydrophobization of cotton fabric using various silanes, including dichlorodimethylsilane, to enhance fabric properties (T Makowski, 2020 - Springer). View Article.




Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
08471CDS025246CDS025245
Quality Level

200

Quality Level

100

Quality Level

-

Quality Level

-

form

liquid

form

liquid

form

-

form

-

mp

-76 °C

mp

-

mp

-

mp

-

bp

70 °C/1013 hPa

bp

-

bp

-

bp

-

storage temp.

2-30°C

storage temp.

-

storage temp.

-

storage temp.

-

density

1.07 g/cm3 at 25 °C

density

-

density

-

density

-


Still not finding the right product?

Explore all of our products under Dichlorodimethylsilane


signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

33.8 °F - closed cup

flash_point_c

1 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters