Skip to Content
Merck
All Photos(1)

Key Documents

810606P

Avanti

16:0 Tempo PC

Avanti Research - A Croda Brand 810606P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phospho(tempo)choline

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€644.00

€644.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€644.00

About This Item

Empirical Formula (Hill Notation):
C48H94N2O9P
CAS Number:
Molecular Weight:
874.24
MDL number:
UNSPSC Code:
41141825
NACRES:
NA.25

€644.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810606P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810606P

lipid type

phospholipids
ESR probes

shipped in

dry ice

storage temp.

−20°C

General description

1,2-dipalmitoyl-sn-glycero-3-phospho(tempo)choline (16:0 Tempo PC) is a spin label[1] phospholipid, where TEMPO moiety is attached to phosphocholine headgroup.[2][3]

Application

1,2-dipalmitoyl-sn-glycero-3-phospho(tempo)choline (16:0 Tempo PC) may be used:
  • as a spin-labeled quencher in liposomes to determine fluorophore depth[4]
  • as spin-labeled phospholipid along with 1,2-dimyristoyl(d54)-sn-glycero-3-phosphocholine-1,1,2,2-d4-N,N,N-trimethyl-d9 (DMPC-d67) to label lidocaine for paramagnetic relaxation measurements[5]
  • as a spin label to investigate the dependence of the packing and polarity of 1,2-dilauroyl-sn-glycero-3-phosphocholine (DLPC) bilayers with temperature or as quenchers of the Laurdan fluorescence to study the Laurdan position in fluid bilayers[1]

Biochem/physiol Actions

1,2-dipalmitoyl-sn-glycero-3-phospho(tempo)choline (16:0 Tempo PC/Tempo-PCSL) can effectively repress Laurdan fluorescence and can also quench the emission of long wavelength state.[1]

Packaging

5 mL Clear Glass Sealed Ampule (810606P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cíntia C De Vequi-Suplicy et al.
Journal of fluorescence, 16(3), 431-439 (2006-06-23)
Laurdan (2-dimethylamino-6-lauroylnaphthalene) is a hydrophobic fluorescent probe widely used in lipid systems. This probe was shown to be highly sensitive to lipid phases, and this sensitivity related to the probe microenvironment polarity and viscosity. In the present study, Laurdan was
Alexander Kyrychenko et al.
The Journal of membrane biology, 253(1), 73-77 (2019-09-22)
The characterization of the behavior of lipid-attached spin probes in a bilayer is of fundamental importance for correct interpretation of the results of both EPR and fluorescence studies of protein-membrane interactions. The knowledge of the immersion depth of TEMPO spin
Victor Vasquez-Montes et al.
Biochimica et biophysica acta. Proteins and proteomics, 1867(7-8), 691-700 (2019-04-21)
Bcl-xL is a member of the Bcl-2 family of apoptotic regulators, responsible for inhibiting the permeabilization of the mitochondrial outer membrane, and a promising anti-cancer target. Bcl-xL exists in the following conformations, each believed to play a role in the
Diego E Sastre et al.
The Journal of biological chemistry, 295(7), 2136-2147 (2019-12-05)
PlsX plays a central role in the coordination of fatty acid and phospholipid biosynthesis in Gram-positive bacteria. PlsX is a peripheral membrane acyltransferase that catalyzes the conversion of acyl-ACP to acyl-phosphate, which is in turn utilized by the polytopic membrane
Nicole Weizenmann et al.
Biochimica et biophysica acta, 1818(12), 3010-3018 (2012-07-31)
The membrane location of the local anesthetics (LA) lidocaine, dibucaine, tetracaine, and procaine hydrochloride as well as their influence on phospholipid bilayers were studied by ³¹P and ¹H magic-angle spinning (MAS) NMR spectroscopy. The ³¹P NMR spectra of the LA/lipid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service