Skip to Content
Merck
All Photos(2)

Key Documents

810609C

Avanti

16:0-18:1 Tempo PC

Avanti Research - A Croda Brand 810609C

Synonym(s):

1-palmitoyl-2-oleoyl-sn-glycero-3-phospho(tempo)choline

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€644.00

€644.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€644.00

About This Item

Empirical Formula (Hill Notation):
C50H96N2O9P
CAS Number:
Molecular Weight:
900.28
MDL number:
UNSPSC Code:
51321705
NACRES:
NA.25

€644.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810609C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810609C

concentration

1 mg/mL (810609C-1mg)

lipid type

phospholipids
ESR probes

shipped in

dry ice

storage temp.

−20°C

General description

16:0-18:1 Tempo PC is a stable isotope and acts as an erythrocyte sedimentation rate (ESR) probe.

Packaging

5 mL Clear Glass Sealed Ampule (810609C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service