Skip to Content
Merck
All Photos(2)

Key Documents

810284P

Avanti

TopFluor Lyso PC

Avanti Research - A Croda Brand 810284P, powder

Synonym(s):

1-(dipyrrometheneboron difluoride)undecanoyl-2-hydroxy-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€231.00

€231.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€231.00

About This Item

Empirical Formula (Hill Notation):
C32H53BF2N3O7P
CAS Number:
Molecular Weight:
671.56
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€231.00


Estimated to ship on31 May 2025


Request a Bulk Order

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

powder

packaging

pkg of 1 × 1 mg (810284P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810284P

shipped in

dry ice

storage temp.

−20°C

General description

Fluorescent derivative of lyso phosphatidylcholine with spectral properties similar to BODIPY-FL.
Phosphatidylcholines (PC) are the building blocks of eukaryotic membranes. It is also found at high levels in Caenorhabditis elegans lipid extracts.[1] PC is an important component of the mucosal layer of the colon.[2] It is a strong bilayer-forming lipid.[3]

Application

TopFluor Lyso PC may be used as a substrate in phospholipase D (PLD) transphosphatidylation assay (thin-layer chromatography).[4]

Biochem/physiol Actions

Phosphatidylcholine (PC) functions as a surfactant within the mucus to form a hydrophobic surface to inhibit bacterial penetrance.[2] It is used to treat fat embolism. Phosphatidylcholine lowers the levels of cholesterol and triglycerides.[5]

Packaging

5 mL Amber Glass Screw Cap Vial (810284P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimizing a ultrahigh pressure liquid chromatography-time of flight-mass spectrometry approach using a novel sub-2 mum core-shell particle for in depth lipidomic profiling of Caenorhabditis elegans
Witting M, et al.
Journal of Chromatography A, 1359(2), 91-99 (2014)
A Molecular Target for an Alcohol Chain-Length Cutoff
Chung HW, et al.
Journal of Molecular Biology, 431(2), 196-209 (2019)
Integrative Medicine - E-Book (2017)
The Membranes of Cells (2016)
Efficacy of injections of phosphatidylcholine into fat deposits-a non-surgical alternative to liposuction in body-contouring
Karl GH, et al.
Indian journal of plastic surgery : official publication of the Association of Plastic Surgeons of India, 38(2), 119-119 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service