Skip to Content
Merck
All Photos(2)

Key Documents

810281P

Avanti

TopFluor PC

1-palmitoyl-2-(dipyrrometheneboron difluoride)undecanoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

1-palmitoyl-2-(dipyrrometheneboron difluoride)undecanoyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€237.00
5 MG
€377.00
10 MG
€537.00

€237.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€237.00
5 MG
€377.00
10 MG
€537.00

About This Item

Empirical Formula (Hill Notation):
C48H83BF2N3O8P
CAS Number:
Molecular Weight:
909.97
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€237.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810281P-1mg)
pkg of 5 × 1 mg (810281P-5mg)
pkg of 5 × 2 mg (810281P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810281P

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylcholines (PC) are the building blocks of eukaryotic membranes. It is also found at high levels in Caenorhabditis elegans lipid extracts.[1] PC is an important component of the mucosal layer of the colon.[2] It is a strong bilayer-forming lipid.[3]

Application

TopFluor PC may be used to test the incorporation of circulating phosphatidylcholine into the myelin sheath.[4] It may also be used in liposome generation.[5]

Biochem/physiol Actions

Phosphatidylcholine (PC) functions as a surfactant within the mucus to form a hydrophobic surface to inhibit bacterial penetrance.[2] It is used to treat fat embolism. Phosphatidylcholine lowers the levels of cholesterol and triglycerides.[6]

Packaging

5 mL Amber Glass Screw Cap Vial (810281P-10mg)
5 mL Amber Glass Screw Cap Vial (810281P-1mg)
5 mL Amber Glass Screw Cap Vial (810281P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Membranes of Cells (2016)
Integrative Medicine - E-Book (2017)
Lysosomal integral membrane protein-2 as a phospholipid receptor revealed by biophysical and cellular studies
Conrad KS, et al.
Nature Communications, 8(1), 1908-1908 (2017)
Optimizing a ultrahigh pressure liquid chromatography-time of flight-mass spectrometry approach using a novel sub-2 mum core-shell particle for in depth lipidomic profiling of Caenorhabditis elegans
Witting M, et al.
Journal of Chromatography A, 1359(1), 91-99 (2014)
Targeting myelin lipid metabolism as a potential therapeutic strategy in a model of CMT1A neuropathy
Fledrich R, et al.
Nature Communications, 9(1), 3025-3025 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service