Skip to Content
Merck
All Photos(2)

Key Documents

810281C

Avanti

TopFluor PC

1-palmitoyl-2-(dipyrrometheneboron difluoride)undecanoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-palmitoyl-2-(dipyrrometheneboron difluoride)undecanoyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€237.00
5 MG
€377.00
10 MG
€537.00

€237.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€237.00
5 MG
€377.00
10 MG
€537.00

About This Item

Empirical Formula (Hill Notation):
C48H83BF2N3O8P
CAS Number:
Molecular Weight:
909.97
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€237.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810281C-1mg)
pkg of 5 × 1 mL (810281C-5mg)
pkg of 5 × 2 mL (810281C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810281C

concentration

1 mg/mL (810281C-10mg)
1 mg/mL (810281C-1mg)
1 mg/mL (810281C-5mg)

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylcholines (PC) are the building blocks of eukaryotic membranes.[1] PC is an important component of the mucosal layer of the colon[2] and is a strong bilayer-forming lipid.[3]

Application

TopFluor PC has been used as a component in the for fluorescent studies using small unilamellar vesicles.[4] It has also been used in the preparation of unilamellar phospholipid liposomes.[5]

Biochem/physiol Actions

Phosphatidylcholine (PC) functions as a surfactant within the mucus to form a hydrophobic surface to inhibit bacterial penetrance.[2] It is used to treat fat embolism.[6]

Packaging

5 mL Amber Glass Screw Cap Vial (810281C-10mg)
5 mL Amber Glass Screw Cap Vial (810281C-1mg)
5 mL Amber Glass Screw Cap Vial (810281C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The
Integrative Medicine - E-Book (2017)
The
The Membranes of Cells (2016)
Mammalian nonmuscle myosin II binds to anionic phospholipids with concomitant dissociation of the regulatory light chain
Liu X, et al.
The Journal of Biological Chemistry, 291(48), 24828-24837 (2016)
Formation of supported lipid bilayers containing phase-segregated domains and their interaction with gold nanoparticles
Melby ES, et al.
Environmental science. Nano, 3(1), 45-55 (2016)
Optimizing a ultrahigh pressure liquid chromatography-time of flight-mass spectrometry approach using a novel sub-2 mum core-shell particle for in depth lipidomic profiling of Caenorhabditis elegans
Witting M, et al.
Journal of Chromatography A, 1359, 91-99 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service