Skip to Content
Merck
All Photos(1)

Key Documents

800821C

Avanti

18:0-16:0 DG

1-octadecanoyl-2-hexadecanoyl-sn-glycerol, chloroform

Synonym(s):

1-stearoyl-2-palmitoyl-sn-glycerol; DG(18:0/16:0/0:0); 110883

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€161.00

€161.00


Estimated to ship on18 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€161.00

About This Item

Empirical Formula (Hill Notation):
C37H72O5
CAS Number:
Molecular Weight:
596.96
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€161.00


Estimated to ship on18 April 2025


Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 5 mL (800821C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 800821C

concentration

1 mg/mL (800821C-5mg)

lipid type

neutral glycerides
neutral lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O(C(COC(=O)CCCCCCCCCCCCCCCCC)CO)C(=O)CCCCCCCCCCCCCCC

InChI

1S/C37H72O5/c1-3-5-7-9-11-13-15-17-18-20-21-23-25-27-29-31-36(39)41-34-35(33-38)42-37(40)32-30-28-26-24-22-19-16-14-12-10-8-6-4-2/h35,38H,3-34H2,1-2H3

InChI key

VYQDALBEQRZDPL-UHFFFAOYSA-N

General description

18:0-16:0 DG, also called 1-stearoyl-2 palmitoyl-sn-glycerol diacylglycerol (DAG), comprises a glycerol backbone with two carboxylic acids covalently linked to the sn-1 and sn-2 position.[1]

Application

18:0-16:0 DG may be used in the standard curve generation for liquid chromatography-mass spectrometry (LC-MS) analysis.[2]

Biochem/physiol Actions

18:0-16:0 DG, also called 34:0-DG, is phosphorylated by diacylglycerol kinase δ at high glucose levels. DG participates in protein kinase C activation and is a second messenger for many signaling events.[2]

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (800821C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Constanze Müller et al.
Rapid communications in mass spectrometry : RCM, 28(15), 1735-1744 (2014-07-01)
The ionization of neutral diacylglycerols (DAGs) by electrospray ionization mass spectrometry (ESI-MS) is challenging compared with other lipid classes which possess ionic head group conjugations. Although ESI-MS is the method of choice in lipidomic analysis, it is questionable whether all
Dimer ion formation and intermolecular fragmentation of 1, 2-diacylglycerols revealed by electrospray ionization Fourier transform ion cyclotron resonance mass spectrometry for more comprehensive lipid analysis
Muller C, et al.
Rapid Communications in Mass Spectrometry, 28(15), 1735-1744 (2014)
Chiaki Murakami et al.
Analytical biochemistry, 526, 43-49 (2017-03-21)
Phosphatidylcholine (PC)-specific phospholipase C (PC-PLC) hydrolyzes PC to generate the second messenger 1,2-diacylglycerol (DG) and phosphocholine. PC-PLC plays pivotal roles in inflammation, carcinogenesis, tumor progression, atherogenesis, and subarachnoid hemorrhage. Although the activity of PC-PLC in mammalian tissues was discovered approximately

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service