Skip to Content
Merck
All Photos(1)

Key Documents

792574C

Avanti

16:0-18:1 Phosphatidylethanol-IsoPure

Avanti Research - A Croda Brand

Synonym(s):

1-palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanol (sodium salt)-IsoPure

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€187.00

€187.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€187.00

About This Item

Empirical Formula (Hill Notation):
C39H74NaO8P
CAS Number:
Molecular Weight:
724.96
UNSPSC Code:
51191904
NACRES:
NA.25

€187.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

chloroform solution

packaging

package of 1 × 1 mg

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C39H75O8P.Na/c1-4-7-9-11-13-15-17-19-20-22-24-26-28-30-32-34-39(41)47-37(36-46-48(42,43)45-6-3)35-44-38(40)33-31-29-27-25-23-21-18-16-14-12-10-8-5-2;/h19-20,37H,4-18,21-36H2,1-3H3,(H,42,43);/q;+1/p-1/b20-19-;/t37-;/m1./s1

InChI key

JFUGSRIBTPUDOW-VYMFHCNISA-M

Packaging

5 mL Clear Glass Sealed Ampule (792574C-10MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service