Skip to Content
Merck
All Photos(1)

Key Documents

791646C

Avanti

18:1-d7 MG

Avanti Research - A Croda Brand 791646C

Synonym(s):

1-oleyol(d7)-rac-glycerol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€225.00

€225.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€225.00

About This Item

Empirical Formula (Hill Notation):
C21H33 D7O4
CAS Number:
Molecular Weight:
363.58
UNSPSC Code:
12352211
NACRES:
NA.25

€225.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (791646C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 791646C

concentration

1 mg/mL (791646C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

1-oleoyl(d7)-rac-glycerol (18:1-d7 MG) is a deuterated form of 18:1-MG, which is also called monoolein.[1]
This deuterium labeled lipid is also available in a formulation of multiple lipid standards. The SPLASH LIPIDOMIX Standard includes all of the major lipid classes at ratios relative to human plasma, allowing one easy internal standard solution to be added to every sample.

Application

18:1-d7 MG or 1-oleoyl(d7)-rac-glycerol has been used as an isotope-labeled lipid standard for identification of lipids in mitochondria and lipid droplets samples using mass spectroscopy.[2] It has also been used as an internal standard in ultra-high-performance supercritical fluid chromatography with quadrupole-time-of-flight mass spectrometry (UHPSFC/QTOF-MS) for quantification of pulmonary artery lipids from porcine.[3]

Biochem/physiol Actions

Monoolein (18:1-MG ) serves as a substrate for the enzyme lipase[1] and is a useful lipid additive for meso crystallization and a stabilizer of the lipidic cubic phase (LCP).[4]

Packaging

5 mL Clear Glass Sealed Ampule (791646C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
LIPIDOMIX is a trademark of Avanti Polar Lipids, LLC
SPLASH is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaohua Guo et al.
Bioorganic & medicinal chemistry letters, 25(17), 3464-3467 (2015-07-23)
Compelling evidence have demonstrated the role of lipase activity in the pathogenicity of Malassezia globosa toward dandruff and seborrheic dermatitis (D/SD). As a representative secreted lipase from M. globosa CBS 7966, Malassezia globosa LIP1 (SMG1) is considered a potential anti-dandruff
Alicia Gil-Ramirez et al.
Journal of chromatography. A, 1592, 173-182 (2019-02-03)
Pulmonary artery grafts are needed as cardiovascular bioprosthetics. For successful tissue recellularization after transplantation, lipids have to be removed from the donor artery. Developing a selective process to remove lipids without damaging the extracellular matrix greatly depends on knowing the
Isoprenoid-chained lipid beta-XylOC16+ 4?A novel molecule for in meso membrane protein crystallization
Borshchevskiy V, et al.
Journal of Crystal Growth, 312(22), 3326-3330 (2010)
Tong Liu et al.
Expert review of proteomics, 17(3), 233-242 (2020-04-21)
Background: Colorectal cancer (CRC) represents a third leading cause of cancer-related death worldwide. The reliable diagnostic biomarkers for detecting CRC at early stage is critical for decreasing the mortality.Method: A conjunctive lipidomic approach was employed to investigate the differences in
Florian Geltinger et al.
GeroScience, 42(1), 19-38 (2019-11-05)
Originally Lipid droplets (LDs) were considered as being droplets for lipid storage only. Increasing evidence, however, demonstrates that LDs fulfill a pleiotropy of additional functions. Among them is the modulation of protein as well as lipid homeostasis. Under unfavorable pro-oxidative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service