Skip to Content
Merck
All Photos(1)

Key Documents

791639C

Avanti

15:0-18:1-d7-PS

Avanti Research - A Croda Brand

Synonym(s):

1-pentadecanoyl-2-oleoyl(d7)-sn-glycero-3-phospho-L-serine (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€639.00

€639.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€639.00

About This Item

Empirical Formula (Hill Notation):
C39H66 D7 NNaO10 P
CAS Number:
Molecular Weight:
777.00
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€639.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (791639C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL (791639C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

1-pentadecanoyl-2-oleoyl(d7)-sn-glycero-3-phospho-L-serine (sodium salt) is a is a deuterated phospholipid. The lipid comprises phosphatidylserine attached to pentadecanoic acid and oleic acid. Oleic acid harbors the deuterium ions. Phosphatidylserine is a component of inner cell membrane and is present in metabolically active sites.[1] It is enriched in myelin sheath and is an acidic phospholipid.[2]
This deuterium labeled lipid is also available in a formulation of multiple lipid standards. The SPLASH LIPIDOMIX Standard includes all of the major lipid classes at ratios relative to human plasma, allowing one easy internal standard solution to be added to every sample.

Application

15:0-18:1-d7-PS (1-pentadecanoyl-2-oleoyl(d7)-sn-glycero-3-phospho-L-serine (sodium salt)) has been used:
  • as a component of extraction solvent for lipid isolation from biological samples[3]
  • as an internal standard to spike cell lysate and to quantify lipid species using fourier transform mass spectrometry (FTMS) analysis[4]
  • as an internal standard to spike liver homogenate to quantify lipids by shotgun-electrospray ionization tandem mass spectrometry (ESI-MS)[5]

Biochem/physiol Actions

Phosphatidylserine (PS) coordinates many regulatory functions and is a modulator of enzymes, channels and various receptors. PS supplementation provides relief from exercise induced stress.[1] PS is essential for neurotransmission, prevents nerve cell deterioration and supports cognitive functionality.[2]

Packaging

2 mL Amber Glass Sealed Ampule (791639C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
LIPIDOMIX is a trademark of Avanti Polar Lipids, LLC
SPLASH is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael A Starks et al.
Journal of the International Society of Sports Nutrition, 5, 11-11 (2008-07-30)
Previous research has indicated that phosphatidylserine (PS) supplementation has the potential to attenuate the serum cortisol response to acute exercise stress. Equivocal findings suggest that this effect might be dose dependent. This study aimed to examine the influence of short-term
Impact of plant polyphenols on triacylglycerols and phos-pholipids in rat liver fed with high fat diet
Ma HF, et al.
OIL CROP SCIENCE, 4, 251-264 (2017)
Tong Liu et al.
Expert review of proteomics, 17(3), 233-242 (2020-04-21)
Background: Colorectal cancer (CRC) represents a third leading cause of cancer-related death worldwide. The reliable diagnostic biomarkers for detecting CRC at early stage is critical for decreasing the mortality.Method: A conjunctive lipidomic approach was employed to investigate the differences in
Chuchu Wang et al.
Nature communications, 11(1), 1531-1531 (2020-03-27)
Vesicle associated membrane protein 2 (VAMP2/synaptobrevin2), a core SNARE protein residing on synaptic vesicles (SVs), forms helix bundles with syntaxin-1 and SNAP25 for the SNARE assembly. Prior to the SNARE assembly, the structure of VAMP2 is unclear. Here, by using
Michael J Glade et al.
Nutrition (Burbank, Los Angeles County, Calif.), 31(6), 781-786 (2015-05-03)
The aim of this study was to assess the roles and importance of phosphatidylserine (PS), an endogenous phospholipid and dietary nutrient, in human brain biochemistry, physiology, and function. A scientific literature search was conducted on MEDLINE for relevant articles regarding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service