Skip to Content
Merck
All Photos(1)

Key Documents

710334P

Avanti

18:0 Cardiolipin

Avanti Research - A Croda Brand, powder

Synonym(s):

1′,3′-bis[1,2-Distearoyl-sn-glycero-3-phospho]-glycerol, sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€198.00

€198.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€198.00

About This Item

Empirical Formula (Hill Notation):
C81H156Na2O17P2
CAS Number:
Molecular Weight:
1510.02
UNSPSC Code:
51191904
NACRES:
NA.25

€198.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (710334P-25MG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=P([O-])(OC[C@]([H])(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O)OCC([H])(O)COP([O-])(OC[C@]([H])(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O)=O.[Na+].[Na+]

InChI

1S/C81H158O17P2.2Na/c1-5-9-13-17-21-25-29-33-37-41-45-49-53-57-61-65-78(83)91-71-76(97-80(85)67-63-59-55-51-47-43-39-35-31-27-23-19-15-11-7-3)73-95-99(87,88)93-69-75(82)70-94-100(89,90)96-74-77(98-81(86)68-64-60-56-52-48-44-40-36-32-28-24-20-16-12-8-4)72-

InChI key

MZVKSYZWSXVEFZ-PNVJLZMSSA-L

General description

Cardiolipin (CL) is a polyglycerophospholipid. In eukaryotes, CL is exclusively present in the inner mitochondrial membrane.[1] CL species are complex in structure due to variations in the chain length, degree of unsaturation, as well as the permutations of four fatty acid substituents.[2]

Biochem/physiol Actions

Cardiolipin (CL) is synthesized in mitochondria. CL is essential for mitochondrial bioenergetics and signaling.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (710334P-25MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The phospholipase iPLA2gamma is a major mediator releasing oxidized aliphatic chains from cardiolipin, integrating mitochondrial bioenergetics and signaling
Liu GY, et al.
The Journal of Biological Chemistry, 292(25), 10672-10684 (2017)
Characterization of cardiolipin as the sodiated ions by positive-ion electrospray ionization with multiple stage quadrupole ion-trap mass spectrometry
Hsu FF and Turk J
Journal of the American Society For Mass Spectrometry, 17(8), 1146-1157 (2006)
Oswald Quehenberger et al.
Journal of lipid research, 51(11), 3299-3305 (2010-07-31)
The focus of the present study was to define the human plasma lipidome and to establish novel analytical methodologies to quantify the large spectrum of plasma lipids. Partial lipid analysis is now a regular part of every patient's blood test

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service