Skip to Content
Merck
All Photos(1)

Key Documents

700148P

Avanti

sitosterol-d7

Avanti Research - A Croda Brand 700148P, powder

Synonym(s):

(24-ethyl)-heptadeuteriostigmast-5-en-3β-ol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€377.00

€377.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€377.00

About This Item

Empirical Formula (Hill Notation):
C29H43D7O
CAS Number:
Molecular Weight:
421.75
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€377.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700148P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700148P

shipped in

dry ice

storage temp.

−20°C

General description

Sitosterol-d7 is a deuterated sterol.[1] Sitosterol is distributed widely in plants. It regulates the embryonic phase in plants.[2] β-sitosterol biosynthesis occurs via mevalonate and deoxyxylulose pathways.[3]

Application

Sitosterol-d7 has been used as a deuterated standard in liquid chromatography multiple reaction monitoring (LC-MRM) analysis for the quantification of pollen sterols.[1]

Biochem/physiol Actions

Sitosterol possesses hypocholesterolemic activity. In addition, it also exhibits anticancer activity in colon carcinoma cells (HT-29).[3] D7-sitosterol is used in spiking ATP binding cassette half-transporter knockout mice to understand sitosterol clearance.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (700148P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The story of beta-sitosterol-a review
Saeidnia S, et al.
European Journal of Medicinal Plants, 590-609 (2014)
Erick J Dufourc
Plant signaling & behavior, 3(2), 133-134 (2008-02-01)
Membranes of composition approaching those found in "rafts" of plants, fungi and mammals were investigated by means of solidstate (2)H-NMR, using deuterated dipalmitoyl-phosphatidylcholine ((2)H-DPPC) as a reporter. The dynamics of such membranes was determined through measuring of membrane ordering or
Priyadarshini Chakrabarti et al.
Metabolomics : Official journal of the Metabolomic Society, 15(10), 127-127 (2019-09-21)
Significant annual honey bee colony losses have been reported in the USA and across the world over the past years. Malnutrition is one among several causative factors for such declines. Optimal nutrition serves as the first line of defense against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service