Skip to Content
Merck
All Photos(1)

Key Documents

700094P

Avanti

cholestanol-d5

Avanti Research - A Croda Brand

Synonym(s):

111020

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€377.00

€377.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€377.00

About This Item

Empirical Formula (Hill Notation):
C27H43D5O
CAS Number:
Molecular Weight:
393.70
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€377.00


Check Cart for Availability

Request a Bulk Order

description

2,2,3,4,4-pentadeuterio-5α-cholestan-3β-ol

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700094P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@]1(C([C@H]2[C@@]([C@@H]3[C@H]([C@H]4[C@@]([C@H](CC4)[C@@H](CCCC(C)C)C)(CC3)C)CC2)(CC1([2H])[2H])C)([2H])[2H])[2H]

InChI key

QYIXCDOBOSTCEI-PFPUTWTGSA-N

General description

Cholestanol-d5 is a deuterated form of cholestanol. Cholestanol is a plant saturated sterol and has Δ5 double bond to the 5α position in the cholesterol structure[1] and is a 5α-dihydro derivative of cholesterol.[2]

Biochem/physiol Actions

Cholestanol deuterated has been administered in small intestine samples for the quantification of sterols by gas chromatography-mass spectrometry.[1] The levels of cholesterol and cholestanol are elevated in tendons and brain in cerebrotendinous xanthomatosis due to the CYP27A1 (Cytochrome P450 Family 27 Subfamily A Member 1) deficiency.[3] Cholestanol-d5 is useful as an internal standard to spike sediment samples for tandem mass spectrometry (MS/MS) for fecal sterol quantification.[4] Cholestanol is a cholesterol absorption biomarker.[5]

Packaging

5 mL Amber Glass Screw Cap Vial (700094P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B P Lyons et al.
Marine pollution bulletin, 100(2), 689-698 (2015-08-01)
Microbial water quality and concentrations of faecal sterols in sediment have been used to assess the degree of sewage contamination in Kuwait's marine environment. A review of microbial (faecal coliform, faecal streptococci and Escherichia coli) water quality data identified temporal
Ann Båvner et al.
Journal of lipid research, 51(9), 2722-2730 (2010-06-01)
The rare disease cerebrotendinous xanthomatosis (CTX) is due to a lack of sterol 27-hydroxylase (CYP27A1) and is characterized by cholestanol-containing xanthomas in brain and tendons. Mice with the same defect do not develop xanthomas. The driving force in the development
S Shefer et al.
The Journal of clinical investigation, 74(5), 1773-1781 (1984-11-01)
Large amounts of cholestanol, the 5 alpha-dihydro derivative of cholesterol are found in tissues of patients with the rare inherited sterol storage disease cerebrotendinous xanthomatosis. Although small amounts of cholestanol are present in virtually every tissue of normal man, little
Monica Gelzo et al.
Lipids in health and disease, 18(1), 235-235 (2020-01-01)
Cerebrotendinous xanthomatosis (CTX) and Lathosterolosis represent two treatable inherited disorders of cholesterol metabolism that are characterized by the accumulation of cholestanol and lathosterol, respectively. The age of the patients suspected of having these disorders is highly variable due to the
Michael Igel et al.
Journal of lipid research, 44(3), 533-538 (2003-02-04)
The recent identification of the aberrant transport proteins ABCG5 and ABCG8 resulting in sitosterolemia suggests that intestinal uptake of cholesterol is an unselective process, and that discrimination between cholesterol and plant sterols takes place at the level of sterol efflux

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service