Skip to Content
Merck
All Photos(1)

Key Documents

181141C

Avanti

Sunflower Lyso PC

Avanti Research - A Croda Brand

Synonym(s):

1-Linoleoyl-2-hydroxy-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352211
NACRES:
NA.79

biological source

(Sunflower)

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (181141C-25MG)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL

density

1.49 g/cm3

functional group

phospholipid

lipid type

phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OC[C@@H](O)COP([O-])(=O)OCC[N+](C)(C)C

InChI

1S/C24H50NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-24(27)30-21-23(26)22-32-33(28,29)31-20-19-25(2,3)4/h23,26H,5-22H2,1-4H3/t23-/m1/s1

InChI key

ASWBNKHCZGQVJV-HSZRJFAPSA-N

Application

Lipids research

Packaging

5 mL Clear Glass Sealed Ampule (181141C-25MG)

Analysis Note

Average Fatty Acid Distribution: 9.4% - 16:0; 2.8% - 18:0; 19.8% - 18:1; 67.4% - 18:2; 0.1% - 18:3; 0.1% - 20:0; 0.1% - 20:1; 0.3% 22:0

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service