Skip to Content
Merck
All Photos(1)

Key Documents

690050C

Avanti

Soy Phospholipid Mixture

Avanti Research - A Croda Brand

Synonym(s):

Soy Phospholipid Mixture, Phospholipids from Soybean, Contains Soy PC (3.8mg) Soy PE (3.0mg) Soy PI (1.8mg) Soy PA (0.7mg) Soy LPC (0.7mg) with 0.1% BHT

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€209.00

€209.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€209.00

About This Item

UNSPSC Code:
51191904
NACRES:
NA.25

€209.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (HPLC)

form

liquid

packaging

pkg of 1 × 1 mL (690050C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (690050C-10mg)

lipid type

lipid extracts
phospholipids

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Soy phospholipid mixture has been used as a phospholipid standard for lipid extraction.[1]

Packaging

5 mL Clear Glass Sealed Ampule (690050C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seipin performs dissectible functions in promoting lipid droplet biogenesis and regulating droplet morphology
Cartwright BR, et al.
Molecular Biology of the Cell, 26(4), 726-739 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service