Skip to Content
Merck
All Photos(1)

Key Documents

Z743823

ART® Non-Filtered Tip

ART 10, size 10 μL, reload tower, sterile

Synonym(s):

aerosol barrier tips, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip, sterile barrier pipette tip

Sign Into View Organizational & Contract Pricing

Select a Size

5 X 960 EA
€497.00

€497.00


Check Cart for Availability


Select a Size

Change View
5 X 960 EA
€497.00

About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
10 μL
sterility:
sterile
feature:
ART 10
reload tower
packaging:
pkg of 5X960 ea
manufacturer/tradename:
Molecular BioProducts 3501-RT

€497.00


Check Cart for Availability

sterility

sterile

feature

ART 10
reload tower

packaging

pkg of 5X960 ea

manufacturer/tradename

Molecular BioProducts 3501-RT

technique(s)

PCR: suitable

size

10 μL

volume

10 μL

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

ART™ Non-filtered Hinged Rack Pipette Tips, which are made for the way you work, allows you to access tips with just one hand. You need not be concerned about your tips spilling on the bench or floor anymore. In just three simple steps, you can go from hinged to lift-off and maximize your productivity. Just unhinge, turn, and realign the cover to have a conventional lift-off when necessary. These tips are appropriate if contamination is not a concern.

Features and Benefits

  • Complete selection of non-filtered and specialty tips provides ART quality, performance, and reliability.
  • Racks allow saving time by offering quick and easy access to pipette tips.
  • The front latch closes and locks to provide safe and secure storage

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service