Skip to Content
Merck
All Photos(1)

Documents

Z742572

Optional rotor for PlateFuge Microcentrifuge

for 2 x deepwell plates up to 35 mm height

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909
NACRES:
NB.78

feature

to hold 2 x deep well plates (up to 35 mm tall)
for 2 x deepwell plates up to 35 mm height

packaging

pack of 1 ea

manufacturer/tradename

Benchmark Scientific C2000-DWMP

parameter

1500 max. RCF

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models PlateFuge Microcentrifuge

General description

PlateFuge Microplate Optional Rotor

This optional rotor can carry deep well plates up to 35mm tall. This rotor also spins at a lower speed with the increase in tube size.

Legal Information

PlateFuge is a trademark of Benchmark Scientific, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service