Skip to Content
Merck
All Photos(1)

Documents

Z740920

Sigma-Aldrich

Aldrich® Essentials volumetric pipette, class A, meets ASTM E 969

capacity 1.0 mL, tol. 0.006 mL, blue

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

material

borosilicate glass

feature

class A

packaging

pack of 12 ea

manufacturer/tradename

Sigma-Aldrich

tol. measuring range

0.006 mL

capacity

1.0 mL

color

blue

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Borosilicate glass, reusable, volumetric pipets, Calibrated to contain. Permanent white fill mark and capacity mark. Color coded. Meets Class A, ASTM E 969 specifications.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service