Key Documents
Z693332
Norell® Sample Vault NMR tube caps
closed port cap, red, diam. 3 mm
Select a Size
96 EA
€85.00
€85.00
Estimated to ship on01 April 2025
Select a Size
Change View
96 EA
€85.00
About This Item
UNSPSC Code:
41121800
NACRES:
SB.32
€85.00
Estimated to ship on01 April 2025
packaging
pkg of 96 ea
manufacturer/tradename
Norell SVCP-NORLOC-3-96PK
diam.
3 mm
color
red
General description
Sample Vault caps have superior holding and sealing capabilities which eliminate cap/tube failure in your instrument. The patented design incorporates a white band positioned on the NMR tube that aligns with the base of the cap, indicating a properly locked position. The superior Sample Vault cap attaches semi-permanently for multiple use and for critical applications.
Features and Benefits
  • Superior Sample Vault cap attaches semi-permanently for several uses and critical applications.
  • Ideal for cold refrigeration storage.
  • Suitable for cryo-probes and variable temperature studies.
Legal Information
Norell is a registered trademark of Norell, Inc.
Sample Vault is a trademark of Norell, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)