Skip to Content
Merck
All Photos(1)

Documents

Z693359

Norell® Sample Vault NMR tube caps

closed port cap, red, diam. 5 mm

Synonym(s):

NMR Tube Caps

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
SB.32

packaging

pkg of 96 ea

manufacturer/tradename

Norell SVCP-NORLOC-5-96PK

diam.

5 mm

color

red

Looking for similar products? Visit Product Comparison Guide

General description

Sample Vault caps have superior holding and sealing capabilities which eliminate cap/tube failure in your instrument. The patented design incorporates a white band positioned on the NMR tube that aligns with the base of the cap, indicating a properly locked position. The superior Sample Vault cap attaches semi-permanently for multiple use and for critical applications.

Features and Benefits

  • Superior Sample Vault cap attaches semi-permanently for several uses and critical applications.
  • Ideal for cold refrigeration storage.
  • Suitable for cryo-probes and variable temperature studies.

Legal Information

Norell is a registered trademark of Norell, Inc.
Sample Vault is a trademark of Norell, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service