Unfortunately, there is no capacity specification for this item. The weight bearing capacity depends on the flex angle, as the base acts as a counterbalance. With heavier items, the stand is more likely to tip as the angle widens.
Z689890
Aldrich® flexible support system
with base plate, L 457 mm
Select a Size
€517.00
Estimated to ship on18 May 2025
Select a Size
About This Item
€517.00
Estimated to ship on18 May 2025
Recommended Products
L
457 mm
General description
Each system includes: three nickel-plated clamp heads, 2-prong clamp with 75 mm max. grip, 3-prong clamp with 69 mm max. grip and spring clamp with 13 mm max. grip., and one nickel-plated flex arm in choice of 305 mm length or 457 mm length.
Clamp heads with prongs include vinyl and fiberglass covers for high temperature applications. Utility wrench included with each system.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
-
how much weight this clamp can carry in pounds?
1 answer-
Helpful?
-