Z680818
Duran® stirred reactor caps and accessories, for GL 80 stirred reactor systems
impeller type magnetic stirrer.
Synonym(s):
1200382
Select a Size
€248.00
Select a Size
About This Item
€248.00
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
DURAN® 1200382
diam.
62 mm
Looking for similar products? Visit Product Comparison Guide
General description
- The accessories can be autoclaved and are suitable for use in the biological sector
- The agitating elements are exchangeable and use up to 500 rpm is possible
Features and Benefits
- Manufactured from chemically resistant materials: PP, PTFE, PEEK, stainless steel
- Drive with standard magnetic stirrers up to 500 rpm and allows improved mixing compared to stir bars
- The variable stirrer shaft can be vertically adjusted to the optimal position in either 500, 1000 or 2000 ml DURAN® GLS 80® bottles
- A shorter stirrer shaft is available for the 250 ml DURAN® GLS 80® bottles
- The 4x threaded ports (2x GL 14 and 2x GL 18) can be blanked off or used for connecting tubing or measurement probes
- Flexible: The proven DURAN® GL 45 / GLS 80 connection system components (2x GL 14 and 2x GL 18) can be used with 1.6 to 12.0 mm outer diameter semi-rigid tubing to add or remove liquid or gas during the mixing process
- Fully Autoclavable components
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service