Documents
CR0003L200
Millipore
Single-use 3L stirred tank bioreactor (green)
About This Item
UNSPSC Code:
23151806
eCl@ss:
32031690
Quality Level
General description
A single-use, stirred tank bioreactor designed for the non-GMP cultivation of mammalian cell lines. Its rigid design includes a semi-hemispherical shaped polycarbonate vessel, a multi-port high-density polyethylene (HDPE) headplate, a scoping marine impeller, both open pipe and sintered spargers, weldable C-Flex tubing, and all necessary gas inlet and outlet filters. Because the vessel is molded component, additional ports were added below the minimum working volume level including, one luer actuated sample port, one liquid inlet/outlet port, two liquid addition lines, direct sparging lines and a harvest line. All gas and liquid addition ports located below the minimum working volume incorporate check valves to ensure uni-directional flow of gas or liquid. The CellReady 3L Bioreactor is preassembled, gamma irradiated and shipped ready to use.
consumable
Product No.
Description
Pricing
equipment
Product No.
Description
Pricing
Visit the Document Library
Articles
A step-by-step overview of suspension-based, transient transfection bioreactor process development and scaleup of lentivirus production.
This page describes the design attributes of the Cellicon® filter assembly for cell retention and how they simplify scale-up of intensified upstream processes.
This page describes the financial and operational advantages offered by the Cellicon® Cell Retention Device in an N-1 perfusion process at the 2,000 L scale in comparison to a cell retention device from another supplier.
High cell density cryopreservation (HCDC) is a closed processing alternative to the traditional seed train expansion process. Learn more about the benefits of HCDC.
See All
Related Content
Biopharmaceutical developers need efficient, productive upstream processes that deliver high quality monoclonal antibodies (mAbs) and other types of recombinant proteins. Process intensification can future-proof their manufacturing strategies, reduce costs, and increase facility flexibility.
Mobius® iFlex Bioreactor family offers single-use bioreactors for mammalian cell culturing from development to production.
Biopharmaceutical developers need efficient, productive upstream processes that deliver high quality monoclonal antibodies (mAbs) and other types of recombinant proteins. Process intensification can future-proof their manufacturing strategies, reduce costs, and increase facility flexibility.
Learn more on mAB bioreactor production, and the products you need for this step.
See All