Skip to Content
Merck
All Photos(1)

Key Documents

Z679968

Bio-Pure pipetting reservoir

Bio-Pure™ pipetting reservoir
1 of 1 reviewers received a sample product or took part in a promotion

capacity 50 mL

Synonym(s):

multichannel pipette reservoir, reagent reservoir

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€121.00

€121.00


Check Cart for Availability


Select a Size

Change View
100 EA
€121.00

About This Item

UNSPSC Code:
41121522
NACRES:
NB.01

€121.00


Check Cart for Availability

material

PET (Bio-degradable)
V-bottom bottom
white

sterility

sterile

packaging

pack of 100 ea

manufacturer/tradename

Diversified Biotech REBP-5000

capacity

50 mL

Looking for similar products? Visit Product Comparison Guide

General description

Bio-Pure Reagent Reservoirs are made from recaputured PET that does not rely on additional fossil fuel-derived petrochemicals. In addition, reservoirs are sealed in bio-degradable bags and packed in recycled cardboard cases. As a result, Bio-Pure Reservoirs are a more eco-friendly alternative. "Trough within a trough" maximizes reagent recovery. A wide base and reinforced sidewalls avoids spills and provides stability. Gamma irradiated RNASE/DNASE free

Features and Benefits

  • Bio-Pure pipetting reservoirs are γ-irradiated with 50ml capacity.
  • Free of RNase/DNase
  • Constructed from biodegradable PET material for environmentally conscious disposal.
  • Specialized "trough within a trough" design for optimal reagent recovery.
  • Pour-off spouts at all four corners and a wide base for improved stability and spill prevention.
  • Sterilized for safe and contamination-free usage.

Legal Information

Bio-Pure is a trademark of Diversified Biotech

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. San Juan, PR
    • Review 1
    • Votes 0
    5 out of 5 stars.

    Great reservoir!

    Great reservoir! They are easy to use and can handle appropriate volume. Will order again.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service